Skip to Content
Merck
All Photos(1)

Documents

P6740

Sigma-Aldrich

17-Phenyl-tri-norprostaglandin F-ethyl amide

≥95%, solid

Synonym(s):

Bimatoprost

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37NO4
CAS Number:
Molecular Weight:
415.57
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95%

form

solid

storage temp.

−20°C

SMILES string

CCNC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)CCc2ccccc2

InChI

1S/C25H37NO4/c1-2-26-25(30)13-9-4-3-8-12-21-22(24(29)18-23(21)28)17-16-20(27)15-14-19-10-6-5-7-11-19/h3,5-8,10-11,16-17,20-24,27-29H,2,4,9,12-15,18H2,1H3,(H,26,30)/b8-3-,17-16+/t20-,21+,22+,23-,24+/m0/s1

InChI key

AQOKCDNYWBIDND-FTOWTWDKSA-N

Gene Information

human ... PTGFR(5737)

Application

Potent FP prostanoid receptor agonist used in animal models for glaucoma.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsay Machen et al.
Expert opinion on pharmacotherapy, 21(10), 1269-1282 (2020-04-02)
Fixed-combination glaucoma medications have altered the paradigm of ocular hypertension and glaucoma treatment and are in widespread use today. A comprehensive review of fixed-combination medications will help educate and inform providers for optimal patient care. In this review, the authors
Felipe A Medeiros et al.
Ophthalmology, 127(12), 1627-1641 (2020-06-17)
To evaluate the intraocular pressure (IOP)-lowering efficacy and safety of 10- and 15-μg bimatoprost implant in subjects with open-angle glaucoma (OAG) and ocular hypertension (OHT) after initial and repeated administrations. Randomized, 20-month, multicenter, subject- and efficacy evaluator-masked, parallel-group, phase 3
Hong Liang et al.
Molecular vision, 15, 1690-1699 (2009-08-28)
Using an established rabbit toxicological model, this in vivo study compared the ocular cytotoxicity of four topical intraocular pressure (IOP)-lowering agents: the commercial benzalkonium chloride (BAC)-containing solutions of 0.005% latanoprost, 0.004% travoprost, 0.03% bimatoprost (containing 0.02%, 0.015%, and 0.005% BAC
Marta Zezula et al.
Journal of pharmaceutical and biomedical analysis, 174, 348-359 (2019-06-17)
A simple, rapid and accurate ultra-high performance liquid chromatographic (UHPLC) method with a UV detection for the determination of the chemical purity and assay of bimatoprost (BT-1) was developed. The chromatographic separation was achieved with the use of an Acquity
Marilita M Moschos et al.
Drug design, development and therapy, 10, 3977-3981 (2016-12-21)
The aim of this study was to investigate the effect of different prostaglandin analogs on platelet-activating factor (PAF) levels. Three prostaglandin analogs were selected: bimatoprost 0.3 mg/mL, latanoprost 50 μg/mL, and tafluprost 15 μg/mL. Each drug sample was tested for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service