Skip to Content
Merck
All Photos(3)

Key Documents

P6501

Sigma-Aldrich

Phenyl-β-D-galactopyranoside

≥98% (TLC)

Synonym(s):

P-Gal, Phenyl β-D-galactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O6
CAS Number:
Molecular Weight:
256.25
Beilstein:
87518
EC Number:
MDL number:
UNSPSC Code:
41141627
PubChem Substance ID:
NACRES:
NA.52

Assay

≥98% (TLC)

form

powder

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccccc2)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H16O6/c13-6-8-9(14)10(15)11(16)12(18-8)17-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9+,10+,11-,12-/m1/s1

InChI key

NEZJDVYDSZTRFS-YBXAARCKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenyl-β-D-galactopyranoside is used as a substrate for detecting β-galactosidase enzymatic activity.

Application

Phenyl-β-D-galactopyranoside has been used in mutant phage screening.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ring et al.
Biochemical and biophysical research communications, 131(2), 675-680 (1985-09-16)
The pH profiles of beta-galactosidase, having tyr replaced by m-fluorotyrosine, were compared to those of normal enzyme. The inflection point on the alkaline side was lowered about 1.5 pH units in the fluoro-enzyme, corresponding to the difference in the phenolic
Zbynek Oktabec et al.
Molecules (Basel, Switzerland), 15(12), 8973-8987 (2010-12-15)
Mixtures of ibandronate monosodium salt with eleven gluco- and/or galacto-pyranoside derivatives as counterions were designed to prepare co-crystals with improved intestinal absorption. In general, gastrointestinal absorption of bisphosphonates after oral administration is approximately 1%. Co-crystals were generated by means of
Daniel D Mitchell et al.
Bioorganic & medicinal chemistry, 12(5), 907-920 (2004-02-26)
With the aim of developing high-affinity mono and multivalent antagonists of cholera toxin (CT) and Escherichia coli heat-labile enterotoxin (LT) we are using the galactose portion of the natural receptor ganglioside GM1 as an anchoring fragment in structure-based inhibitor design
Ziyu Deng et al.
International journal of biological macromolecules, 137, 69-76 (2019-07-02)
β-Galactosidase (β-Gal) as dietary supplement has the ability to alleviate symptoms of lactose intolerance. This study investigated the ability of oligosaccharides to protect β-Gal against heat stress. Four kinds of oligosaccharides including Isomalto-oligosaccharides (IMO), Xylo-oligosaccharides (XOS), Konjac-oligosaccharides (KOS), and Mycose
Jules A A C Heuberger et al.
The Lancet. Haematology, 4(8), e374-e386 (2017-07-04)
Substances that potentially enhance performance (eg, recombinant human erythropoietin [rHuEPO]) are considered doping and are therefore forbidden in sports; however, the scientific evidence behind doping is frequently weak. We aimed to determine the effects of rHuEPO treatment in well trained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service