Skip to Content
Merck
All Photos(1)

Key Documents

P5390

Sigma-Aldrich

Prostaglandin B2

≥98%, synthetic

Synonym(s):

(5Z,13E,15S)-15-Hydroxy-9-oxoprosta-5,8(12),13-trien-1-oic acid, PGB2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45
Beilstein:
2153006
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98%

form

solution

solubility

ethanol: ~100 mg/mL
PBS, pH 7.2: ~2 mg/mL
DMSO: ~50 mg/mL

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C1=C(C\C=C/CCCC(O)=O)C(=O)CC1

InChI

1S/C20H30O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h4,7,12,14,17,21H,2-3,5-6,8-11,13,15H2,1H3,(H,23,24)/b7-4-,14-12+/t17-/m0/s1

InChI key

PRFXRIUZNKLRHM-HKVRTXJWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Prostaglandin B2 has been used as an internal standard in high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Prostaglandin B2 (PGB2) is an eicosanoid and a lipid mediator. It is synthesized by the dehydration of PGE2. It is a primary prostaglandin associated with the osteoblasts and may induce pulmonary hypertension. PGB2 is also reported to inhibit glucagon stimulated glycogenolysis and forskolin-stimulated adenylate cyclase activity in plasma membranes.

Physical form

Prostaglandin B2 is supplied as a solution in methyl acetate

Preparation Note

PGB2 is provided as a solution in methyl acetate. To change the solvent, evaporate the methyl acetate under a gentle stream of nitrogen and immediately add the solvent of choice. PGB2, purged with an inert gas, is soluble in ethanol at ~100 mg/mL, DMSO at ~50 mg/mL and in dimethyl formamide at ~ 75 mg/mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.8 °F

Flash Point(C)

-9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation of sulfidopeptide leukotrienes by reversed-phase high-performance liquid chromatography.
M Müller et al.
Journal of chromatography, 343(1), 213-218 (1985-09-13)
K Büyükgüzel et al.
Insect biochemistry and molecular biology, 32(4), 435-443 (2002-03-12)
We describe prostaglandin (PG) biosynthesis by isolated midgut preparations from tobacco hornworms, Manduca sexta. Microsomal-enriched midgut preparations yielded four PGs, PGA/B(2), PGD(2), PGE(2) and PGF(2alpha), all of which were confirmed by analysis on gas chromatography--mass spectrometry (GC--MS). PGA and PGB
EXTRACT FROM NETTLE SEEDS (URTICA DIOICA L.) DECREASES THE SYNTHESIS OF LIPOXIN Asub 4 THROUGH INHIBITION OF THE DEVELOPMENT OF FLUORIDE-INDUCED INFLAMMATION IN THP1 MONOCYTES/MACROPHAGES
Lukomska A, et al.
Fluoride null
Sarah F Muldoon et al.
NeuroImage. Clinical, 18, 599-607 (2018-05-31)
Cognitive effort is known to play a role in healthy brain state organization, but little is known about its effects on pathological brain dynamics. When cortical stimulation is used to map functional brain areas prior to surgery, a common unwanted
J H Feyen et al.
Prostaglandins, 28(6), 769-781 (1984-12-01)
The effects of parathyroid hormone (PTH), dihydroxycholecalciferol (1,25-(OH)2 D3), thrombin, epidermal growth factor (EGF) and 12-o-tetradecanoylphorbol-13-acetate (PMA) on the biosynthesis and release of arachidonic acid metabolites were studied in primary cultures of osteoblast-like cells isolated from 18-day-old chick embryo calvaria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service