Skip to Content
Merck
All Photos(1)

Key Documents

P5136

Sigma-Aldrich

Poly-γ-benzyl-L-glutamate

mol wt 150,000-350,000

Synonym(s):

PBLG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

powder

Quality Level

mol wt

150,000-350,000

color

white to light yellow

storage temp.

−20°C

SMILES string

[N+H3][C@@H](CCC(=O)OCc1ccccc1)C(=O)[O-]

InChI

1S/C12H15NO4/c13-10(12(15)16)6-7-11(14)17-8-9-4-2-1-3-5-9/h1-5,10H,6-8,13H2,(H,15,16)/t10-/m0/s1

InChI key

BGGHCRNCRWQABU-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospun Poly(γ-Benzyl-l-Glutamate) and Its Alkali-Treated Meshes: Their Water Wettability and Cell-Adhesion Potential.
Hakamada Y, Ohgushi N, et al.
Journal of Biomaterials Science. Polymer Edition (2011)
Ma Elisa Martínez-Barbosa et al.
Bioconjugate chemistry, 20(8), 1490-1496 (2009-08-19)
In the present work, the possibility to obtain PEGylated nanoparticles from two PBLG derivatives, PEG-b-poly(γ-benzyl L-glutamate), PBLG-PEG-60, and poly(γ-benzyl L-glutamate), PBLG-Bnz-50, by nanoprecipitation has been investigated. Particles were prepared not only from one polymer (PBLG-PEG-60 or PBLG-Bnz-50), but also from
Hana Robson Marsden et al.
Journal of the American Chemical Society, 132(7), 2370-2377 (2010-01-30)
A new class of peptide has been created, polypeptide-b-designed peptides, which unites the useful qualities of the two constituent peptide types. We demonstrate the synthesis and self-assembly possibilities of this class of peptide chimera with a series of amphiphilic polypeptide-b-designed
Rajeswari Ravichandran et al.
Biomaterials, 33(3), 846-855 (2011-11-04)
Tissue engineering and nanotechnology have enabled engineering of nanostructured materials to meet the current challenges in bone treatment owing to rising occurrence of bone diseases, accidental damages and defects. Poly(L-lactic acid)/Poly-benzyl-L-glutamate/Collagen (PLLA/PBLG/Col) scaffolds were fabricated by electrospinning and nanohydroxyapatite (n-HA)
Uday Ramesh Prabhu et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 202(2), 217-222 (2009-12-17)
We report the application of z-COSY experiment and a band selected version of it by employing a selective 90 degrees pulse entitled BASE-z-COSY for precise chiral discrimination, quantification of enantiomeric excess and the analyses of the (1)H NMR spectra of

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service