Skip to Content
Merck
All Photos(2)

Key Documents

P4506

Sigma-Aldrich

Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake)

Type IV, crude dried venom

Synonym(s):

5′-Exonuclease, Oligonucleate 5′-nucleotidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type IV

Quality Level

form

crude dried venom

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Research area: Cell signalingPhosphodiesteraseI (PDEI) exists in three isoforms PDE1A,PDE1B, and PDE1C(2) that are locatedin cardiovascular, central nervous system, and other organs.

Application

Phosphodiesterase I from Crotalus atrox (Western Diamondback Rattlesnake) has been used to analyse its effect on intracellular calcium concentration of neutrophils. It has also been used in combination with other enzymes to determine the activity of phosphatase enzyme.
Phosphodiesterase I, product P4506 from western diamondback rattlesnake, is used in phosphodiesterase activation assays for calmodulin. It is added to hydrolyze AMP.

Biochem/physiol Actions

Phosphodiesterase I breaks phosphodiester bonds and catalyzes the hydrolysis of various nucleotide polyphosphates. Phosphodiesterase I is released from eucaryotic plasma membranes by phosphatidylinositol-specific phospholipase C. Phosphodiesterase I (PDEI) can hydrolyze 3′,5′-cyclicadenosine monophosphate (cAMP) and 3′,5′-cyclic guanosine monophosphate (cGMP).It forms a complex with Ca2+/calmodulin (CaM) that leads to the activationof its catalytic activity.PDEIshows biotherapeutic effects for the treatment of cancer, diseases, neurocognitive,pulmonary, metabolic, heart, renal, neurological disorders, and endocrineabnormalities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Identification of Lys49-PLA2 from crude venom of Crotalus atrox as a human neutrophil-calcium modulating protein
Sultan MD, et al.
The Korean Journal of Physiology & Pharmacology : Official Journal of the Korean Physiological Society and the Korean Society of Pharmacology, 20(2), 177-183 (2016)
Assessment of bioavailable organic phosphorus in tropical forest soils by organic acid extraction and phosphatase hydrolysis
Darch T, et al.
Geoderma, 284, 93-102 (2016)
B Fuss et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(23), 9095-9103 (1997-12-31)
One of the more complex developmental processes occurring postnatally in the CNS is the formation of the myelin sheath by oligodendrocytes. To examine the molecular events that take place during myelination, we isolated oligodendrocyte-derived cDNA clones, one of which (p421.HB)
C J Brokaw et al.
The Journal of cell biology, 100(6), 1875-1883 (1985-06-01)
Sea urchin spermatozoa demembranated with Triton X-100 in the presence of EGTA, termed potentially asymmetric, generate asymmetric bending waves in reactivation solutions containing EGTA. After they are converted to the potentially symmetric condition by extraction with Triton and millimolar Ca++
Omer Murik et al.
The Plant journal : for cell and molecular biology, 101(6), 1269-1286 (2019-10-29)
Mitochondria serve as major sites of ATP production and play key roles in many other metabolic processes that are critical to the cell. As relicts of an ancient bacterial endosymbiont, mitochondria contain their own hereditary material (i.e. mtDNA, or mitogenome)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service