Skip to Content
Merck
All Photos(1)

Key Documents

P3808

Sigma-Aldrich

Paraplast X-TRA®

for tissue embedding

Synonym(s):

Tissue Embedding Media

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

solid

mp

50-54 °C (lit.)

application(s)

hematology
histology

storage temp.

2-8°C

General description

N/A

Application

N/A

Legal Information

Paraplast X-TRA is a registered trademark of Leica Biosystems

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Cristina Kalb et al.
Archives of environmental contamination and toxicology, 70(4), 793-801 (2015-08-08)
The health effects of environmental chemicals on animals and humans are of growing concern. Human epidemiological and animal study data indicate that reproductive disorders and diseases begin early during prenatal and postnatal development. An increase of human male reproductive disturbance
A Combination of Culture Conditions and Gene Expression Analysis Can Be Used to Investigate and Predict hES Cell Differentiation Potential towards Male Gonadal Cells
Kristin Ros
PLoS ONE, 2015 null
GH overexpression decreases spermatic parameters and
reproductive success in two-years-old transgenic zebrafish
males
Animal Reproduction Science, 139 (2013)
In vitro Spermatogenesis ? Optimal Culture Conditions for Testicular Cell Survival, Germ Cell Differentiation, and Steroidogenesis in Rats
Ahmed Reda
Frontiers in Endocrinology, 5 (2014)
Seyedeh Ainaz Shirangi et al.
Fish physiology and biochemistry, 42(6), 1741-1754 (2016-06-28)
The effect of abrupt and 5-day gradual salinity transfers from freshwater (FW) to 11 ‰ Caspian Sea brackish water (BW) was investigated in juvenile Persian sturgeon Acipenser persicus with three different weight groups: 1-2 g (1.62 ± 0.27 g), 2-3 g (2.55 ± 0.41 g) and 3-5 g (4.28 ± 0.76 g). Mortality

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service