Skip to Content
Merck
All Photos(1)

Key Documents

P2980

Sigma-Aldrich

Pertussis toxin

from Bordetella pertussis, buffered aqueous glycerol solution, protein endotoxin

Synonym(s):

Histamine-sensitizing factor, IAP, Islet Activating Protein, PTX, Pertussigen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Pertussis toxin from Bordetella pertussis, buffered aqueous glycerol solution

description

aseptically packaged

Quality Level

form

buffered aqueous glycerol solution

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(C)C(=O)CCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C26H52O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28-2/h3-25H2,1-2H3

InChI key

WOPKHAQDUMDJIY-UHFFFAOYSA-N

Gene Information

Bordetella pertussis Tohama I ... ptxA(2665068) , ptxB(2665069) , ptxC(2665408)

Looking for similar products? Visit Product Comparison Guide

General description

Pertussis (Pertussis) toxin (PTX) is a protein isolated from the bacterium Bordatella (Bordatella) coccobaccillus. The toxin is excreted in a non-active state. When the B oligomer of the bacterium, Bordatella coccobacillus, comes in contact with the cell membrane, an S1 subunit of its A protomer is activated. This activation may be mediated by glutathione and/or ATP.

Application

Pertussis toxin from Bordetella pertussis has been used: to enhance cardiac myosin immunization of rats to induce myocarditis; to break down the blood–brain barrier in the Cuprizone and cuprizone/MOG-EAE model; as an inhibitor of Gαi signaling
The toxin is released from B. pertussis in an inactive form. When the pertussis toxin B oligomer binds to the cell membrane, the S1 subunit of its A protomer becomes activated, perhaps through the action of glutathione and ATP. A protocol for activating pertussis toxin in vitro is given by Kaslow, et al.

Biochem/physiol Actions

Pertussis toxin catalyzes the ADP-ribosylation of the α subunits of the heterotrimeric guanine nucleotide regulatory proteins Gi, Go, and Gt. This prevents the G protein heterotrimers from interacting with receptors, thus blocking their coupling and activation. Since the Gα subunits remain in their GDP-bound, inactive state, they are unable to inactivate adenylyl cyclase or open K+ channels.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Physical form

Aseptically filled solution in 50% glycerol, containing 50 mM TRIS, 10 mM glycine, 0.5 M NaCl, pH 7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antonietta Franco et al.
Journal of molecular and cellular cardiology, 121, 60-68 (2018-07-04)
G-protein receptor kinases (GRKs) regulate adult hearts by modulating inotropic, chronotropic and hypertrophic signaling of 7-transmembrane spanning neurohormone receptors. GRK-mediated desensitization and downregulation of β-adrenergic receptors has been implicated in adult heart failure; GRKs are therefore a promising therapeutic target.
Kevin C Kemp et al.
Acta neuropathologica, 135(6), 907-921 (2018-03-16)
Bone marrow-derived cells are known to infiltrate the adult brain and fuse with cerebellar Purkinje cells. Histological observations that such heterotypic cell fusion events are substantially more frequent following cerebellar injury suggest they could have a role in the protection
Jonathan Hoggatt et al.
Cell, 172(1-2), 191-204 (2017-12-12)
Hematopoietic stem cell transplantation is a potential curative therapy for malignant and nonmalignant diseases. Improving the efficiency of stem cell collection and the quality of the cells acquired can broaden the donor pool and improve patient outcomes. We developed a
Arghavan Jahandideh et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 61(11), 1643-1649 (2020-04-15)
Currently available imaging techniques have limited specificity for the detection of active myocardial inflammation. Aluminum 18F-labeled 1,4,7-triazacyclononane-N,N',N″-triacetic acid conjugated folate (18F-FOL) is a PET tracer targeting folate receptor β (FR-β), which is expressed on activated macrophages at sites of inflammation.
Jason M Conley et al.
The Journal of biological chemistry, 297(1), 100881-100881 (2021-06-19)
GPR17 is a G-protein-coupled receptor (GPCR) implicated in the regulation of glucose metabolism and energy homeostasis. Such evidence is primarily drawn from mouse knockout studies and suggests GPR17 as a potential novel therapeutic target for the treatment of metabolic diseases.

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service