Skip to Content
Merck
All Photos(4)

Key Documents

P2007

Sigma-Aldrich

Palatinose hydrate

≥99% (GC)

Synonym(s):

6-O-α-D-Glucopyranosyl-D-fructose, Isomaltulose hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
Beilstein:
93820
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

sugar beets

Assay

≥99% (GC)

form

powder

sweetness

0.5-0.6 × sucrose

technique(s)

gas chromatography (GC): suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

O.OC[C@H]1O[C@H](OC[C@H]2OC(O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.H2O/c13-1-4-6(15)8(17)9(18)11(22-4)21-2-5-7(16)10(19)12(20,3-14)23-5;/h4-11,13-20H,1-3H2;1H2/t4-,5-,6-,7-,8+,9-,10+,11+,12?;/m1./s1

InChI key

XZKUCJJNNDINKX-HGLHLWFZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Palatinose is more slowly absorbed than sucrose and is therefore useful as a sweetener for diabetic patients

Application

Palatinose has been used in a study to assess human intestinal disaccharidases and hereditary disaccharide intolerance. It has also been used in a study to investigate the stimulation of sucrose degradation and starch synthesis in growing potato tubers.

Biochem/physiol Actions

Palatinose is a disaccharide having an α(1→6) linkage between D-glucose and D-fructose and is similar to sucrose in its physicochemical properties. It is produced from sucrose in some bacteria by the action of sucrose isomerase. In mammalian gut, palatinose is hydrolyzed and absorbed much more slowly than sucrose.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoine Fort et al.
Plant physiology, 180(1), 109-123 (2019-02-14)
Green macroalgae of the genus Ulva play a key role in coastal ecosystems and are of increasing commercial importance. However, physiological differences between strains and species have yet to be described in detail. Furthermore, the strains of Ulva used in
K Kawai et al.
Endocrinologia japonica, 32(6), 933-936 (1985-12-01)
Changes in plasma glucose and insulin concentration in response to palatinose ingestion were compared with those to sucrose in eight normal volunteers. When 50 g of palatinose was administered, the plasma glucose gradually increased to its peak of 110.9 +/-
Human intestinal disaccharidases and hereditary disaccharide intolerance. The hydrolysis of sucrose, isomaltose, palatinose (isomaltulose), and a 1,6-alpha-oligosaccharide (isomalto-oligosaccharide) preparation.
A DAHLQVIST et al.
The Journal of clinical investigation, 42, 556-562 (1963-04-01)
A R Fernie et al.
Plant physiology, 125(4), 1967-1977 (2001-04-12)
In the present paper we investigated the effect of the sucrose (Suc) analog palatinose on potato (Solanum tuberosum) tuber metabolism. In freshly cut discs of growing potato tubers, addition of 5 mM palatinose altered the metabolism of exogenously supplied [U-14C]Suc.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service