Skip to Content
Merck
All Photos(4)

Key Documents

P1880

Sigma-Aldrich

Sodium propionate

≥99.0%

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein:
3566934
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0%

form

powder

mp

285-286 °C (lit.)

solubility

H2O: 5 g/5mL

lipid type

unsaturated FAs

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Propionate is a short chain fatty acid, which is a major part of the luminal contents of the colon. It is associated with anti-inflammatory properties.

Application

Sodium propionate has been used as a supplement in transgalacto-oligosaccharides (TOS) medium for the isolation of bifidobacteria. It has been used as a supplement in SCFA (short-chain fatty acid) diet prepared for mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Short-Chain Fatty Acids Protect Against High-Fat Diet-Induced Obesity via a PPAR?-Dependent Switch From Lipogenesis to Fat Oxidation.
den Besten G
Diabetes, 64, 2398-2398 (2015)
Culture-independent analysis of probiotic products by denaturing gradient gel electrophoresis.
Temmerman R
Applied and Environmental Microbiology, 69, 220-220 (2003)
Identification and antibiotic susceptibility of bacterial isolates from probiotic products.
Temmerman R
International Journal of Food Microbiology, 81, 1-1 (2003)
Bingbing Cheng et al.
PloS one, 13(5), e0197380-e0197380 (2018-05-11)
Treatment of prosthetic joint infections often involves multiple surgeries and prolonged antibiotic administration, resulting in a significant burden to patients and the healthcare system. We are exploring a non-invasive method to eradicate biofilm on metal implants utilizing high-frequency alternating magnetic
Anti-inflammatory properties of the short-chain fatty acids acetate and propionate: a study with relevance to inflammatory bowel disease.
Tedelind S
World Journal of Gastroenterology, 13, 2826-2826 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service