Skip to Content
Merck
All Photos(1)

Documents

P0493

Sigma-Aldrich

Monoclonal Anti-PRMT5 antibody produced in mouse

~2 mg/mL, clone PRMT5-21, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Protein-Arginine Methyl Transferase 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PRMT5-21, monoclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa by SDS-PAGE

species reactivity

mouse, hamster, bovine, canine, monkey, rat, human, chicken

concentration

~2 mg/mL

technique(s)

indirect ELISA: suitable
microarray: suitable
western blot: 2 μg/mL using whole cell extract of human melanoma cell line G361

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRMT5(10419)

Related Categories

General description

Monoclonal Anti-PRMT5 (mouse IgG2a isotype) is derived from the PRMT5-21 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with GST-PRMT5 containing the C-terminal region of the human PRMT5. Protein arginine methyltransferases (PRMT) 5 belongs to the type II arginine methyltransferases. PRMT5 (also known as Skb1Hs/janus kinase-binding protein 1 (JBP1) exists as homo-oligomeric complexes, which includes a dimer and tetramer. PRMT5 is found mainly in the cytoplasm. This gene is located on human chromosome 14q11.2.

Immunogen

GST-PRMT5 containing the C-terminal region of the human PRMT5 (amino acids 315-637).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Monoclonal Anti-PRMT5 antibody produced in mouse has been used in enzyme-linked immunosorbent assay (ELISA) and immunoblotting.

Biochem/physiol Actions

Protein arginine methyltransferase 5 (PRMT5) participates in the symmetrical dimethylation of arginine residues. Homooligomerization of PRMT5 has an important role in its ability to methylate the myelin basic protein (MBP) protein. PRMT5 also binds the Janus kinases and are involved in an interferon-signaling pathway. It also helps to produce symmetrical ω-NG-dimethylarginine.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service