Skip to Content
Merck
All Photos(1)

Key Documents

P0107

Sigma-Aldrich

Protease from Rhizopus sp.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

description

acidic protease

form

powder

specific activity

≥0.2 unit/mg solid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Exhibits both proteolytic and lipolytic activities.
Proteases are proteolytic enzymes that digests the proteins. It is classified into acidic, neutral and alkaline proteases based on their pH optimum. Proteases finds its applications in textile industries, laundry and healthcare. It is also being used in food processing industries such as baking and brewing. Rhizopus chinensis is the key species in the production of aspartate proteinase.
Stable in the acid range of pH 3-5. pH Optimum is 3.0.

Application

Protease from Rhizopus sp. has been used in the enzyme treatment of planted and nonplanted scots pine seedlings.
Protease from Rhizopus spp. Has been used in a study to assess the amino acid sequences near the amino termini using automated Edman degradation. It has also been used in a study to investigate inactivation of the enzyme by reaction with diazoacetyl-DL-norleucine methyl ester in the presence of cupric acetate.

Biochem/physiol Actions

Protease from Rhizopus chinensis can be inhibited by pepstatin.

Unit Definition

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 3.0 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Physical form

Supplied as a powder containing dextrin as a stabilizer

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rice bran as a substrate for proteolytic enzyme production
Sumantha A, et al.
Brazilian Archives of Biology and Technology, 49(5), 843-851 (2006)
Stimulation of soil organic nitrogen pool: The effect of plant and soil organic matter degrading enzymes
Kieloaho AJ, et al.
Soil Biology and Biochemistry, 96, 97-106 (2016)
The structure and function of acid proteases. Specific inactivation of an acid protease from Rhizopus chinensis by diazoacetyl-DL-norleucine methyl ester.
F Mizobe et al.
Journal of biochemistry, 73(1), 61-68 (1973-01-01)
J Marciniszyn et al.
The Journal of biological chemistry, 251(22), 7088-7094 (1976-11-25)
Four derivatives of pepstatin, each of which contains the unusual amino acid 4-amino-3-hydroxy-6-methylheptanoic acid (statine) have been prepared. All four are porcine pepsin inhibitors. Both N-acetylstatine and N-acetyl-alanyl-statine are competitive inhibitors for pepsin with Ki values of 1.2 X 10(-4)
Characterization of proteases from Rhizopus species after growth on soybean protein
Heskamp ML and Barz W
Zeitschrift fur Naturforschung C, 52(9-10), 595-604 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service