Skip to Content
Merck
All Photos(1)

Documents

NA0160

Sigma-Aldrich

GenElute HP Plasmid Miniprep Kit

greener alternative

sufficient for 350 preparations

Synonym(s):

GenElute HP Plasmid Kit, HP miniprep kit, fast miniprep kit, fast plasmid isolation kit, plasmid extraction kit, plasmid isolation kit, plasmid purification kit, ultrafast plasmid isolation kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 350 preparations

Quality Level

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

DNA extraction: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

An overnight recombinant E. coli culture is harvested with centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are then removed by a vacuum or spin wash steps. Finally, the bound plasmid DNA is eluted in water or Tris-EDTA buffer.

The GenElute HP Plasmid Miniprep Kit offers an ultrafast and efficient solution for plasmid preparation from E. coli cultures. This kit combines silica-based membrane technology and the convenience of spin or vacuum format,up to 25 μg of high copy plasmid DNA can be recovered from 1-5 mL of E. coli culture in less than 30 minutes. Note that actual yield and optimum volume of culture to use depend on the plasmid and the culture medium.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Application

The recovered plasmid DNA is ready for immediate use in applications such as restriction enzyme digestion, cloning, PCR, transcription, and sequencing.

Features and Benefits

  • Purified plasmid DNA in less than 30 minutes
  • Up to 25 μg of high-copy plasmid DNA
  • Flexibility of a vacuum or spin format
  • No phenol/chloroform extraction or alcohol precipitation required

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of Chemokine Receptor CXCR2 Interacting Proteins Using a Proteomics Approach to Define the CXCR2 ?Chemosynapse?
Dayanidhi Raman
Methods in Enzymology (2009)
Development of Normalized Cdna Library from Fusarium Wilt Infected Roots of a Tolerant Banana Genotype'Calcutta-4'Musa acuminata ssp. burmannicoides
Swarupa V
Journal of Horticultural Science (2014)
Dayanidhi Raman et al.
Methods in enzymology, 460, 315-330 (2009-05-19)
Chemokine-receptor signaling is initiated upon ligand binding to the receptor and continues through the process of endocytic trafficking by the association of a variety of adaptor proteins with the chemokine receptor. In order to define the adaptor proteins that associate
Célia Deville et al.
Cell reports, 27(12), 3433-3446 (2019-06-20)
AAA+ proteins form asymmetric hexameric rings that hydrolyze ATP and thread substrate proteins through a central channel via mobile substrate-binding pore loops. Understanding how ATPase and threading activities are regulated and intertwined is key to understanding the AAA+ protein mechanism.
A Gupta et al.
Genes and immunity, 17(2), 105-109 (2016-01-08)
SLC44A4 is one of the seven novel susceptibility genes that were discovered in the first ever genome-wide association study (GWAS) on ulcerative colitis (UC) in the genetically distinct north Indians. This gene seems to be functionally relevant to disease biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service