N6875
2-Naphthyl acetate
Synonym(s):
β-Naphthyl acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CO2C10H7
CAS Number:
Molecular Weight:
186.21
Beilstein:
1868291
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
form
powder
impurities
<0.2% Free naphthol
mp
67-70 °C (lit.)
solubility
ethanol: 50 mg/mL
storage temp.
−20°C
SMILES string
CC(=O)Oc1ccc2ccccc2c1
InChI
1S/C12H10O2/c1-9(13)14-12-7-6-10-4-2-3-5-11(10)8-12/h2-8H,1H3
InChI key
RJNPPEUAJCEUPV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2-Naphthyl acetate has been used as a substrate:
- in substrate sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) to examine the composition of lipases in larvae and adult specimens
- in the preparation of zymograms to identify proteins with lipase activity
- to measure non-specific esterase activity and β-esterase enzyme activity
Biochem/physiol Actions
2-Naphthyl acetate/β-naphthyl acetate (BNA) is an ester substrate. It can be used in staining gels to detect esterase activity. BNA dissolved in acetone and ethanol produces a dark and very light/no background respectively. Hence destaining is not necessary if ethanol is used to dissolve BNA for esterase staining of gels.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marina Lidwine Olé Sangba et al.
Parasites & vectors, 10(1), 23-23 (2017-01-15)
Knowledge of insecticide resistance status in the main malaria vectors is an essential component of effective malaria vector control. This study presents the first evaluation of the status of insecticide resistance in Anopheles gambiae populations from Bangui, the Central African
β-Naphthyl Acetate in Acetone Produces a Dark Background for Staining of Esterase Activity on Gel.
Deepak Kumar et al.
Indian journal of clinical biochemistry : IJCB, 34(2), 240-242 (2019-05-17)
Digestive lipase activity through development and after fasting and re-feeding in the whiteleg shrimp Penaeus vannamei
Rivera P, et al.
Aquaculture (Amsterdam, Netherlands), 300, 163-168 (2010)
Ewelina Sobstyl et al.
Journal of chromatography. A, 1618, 460942-460942 (2020-02-15)
The fruit of Schisandra chinensis (Chinese magnolia vine), the medicinal plant well-known in Traditional Chinese Medicine, gains great popularity in the modern phytopharmacology. This phenomena is related to the wide and powerful healing properties, including supporting immune, nervous and digestive
Karine da Silva Carvalho et al.
Parasites & vectors, 11(1), 673-673 (2018-12-31)
Bacillus thuringiensis svar. israelensis (Bti) is an effective and safe biolarvicide to control Aedes aegypti. Its mode of action based on four protoxins disfavors resistance; however, control in endemic areas that display high mosquito infestation throughout the year requires continuous
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service