Skip to Content
Merck
All Photos(2)

Key Documents

N5885

Sigma-Aldrich

4-Nitrophenyl α-D-maltoside

glycosidase substrate

Synonym(s):

4-Nitrophenyl a-D-maltopyranoside, 4-Nitrophenyl alpha-D-maltoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO13
CAS Number:
Molecular Weight:
463.39
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥99% (TLC)

form

powder

solubility

water: 49.00-51.00 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](O[C@@]2([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](CO)O[C@@H]1OC3=CC=C([N+]([O-])=O)C=C3

InChI

1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2

InChI key

IAYJZWFYUSNIPN-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I S Lukomskaya et al.
Clinica chimica acta; international journal of clinical chemistry, 244(2), 145-154 (1996-01-31)
The method of assay of neutral alpha-glucosidase from human kidney and urine using beta-maltosides (p-nitrophenyl-beta-D-maltoside [NP-beta-D-maltoside], 2-chloro-4-nitrophenyl-beta-D-maltoside]) [CNP-beta-D-maltoside] and 4-methylumbelliferyl-beta-D-maltosides ([MU-beta-D-maltoside]) as substrates and beta-glucosidase as an auxiliary enzyme is proposed. All three beta-maltosides are suitable substrates for the determination
C C Tseng et al.
Archives of oral biology, 44(2), 119-127 (1999-04-17)
Human salivary alpha-amylase participates in the initial digestion of starch and may be involved in the colonization of viridans streptococci in the mouth. To elucidate the role of histidine residues located near the starch-binding site on the streptococcal-binding activity, the
M Reyes et al.
Journal of bacteriology, 165(3), 918-922 (1986-03-01)
In wild-type Escherichia coli the activity of the maltose transport system is dependent on a periplasmic maltose-binding protein. It has been possible, however, to isolate mutants in which transport activity is mediated by the membrane components of the system and
Reference values for alpha-amylase activity in serum and urine by a new chromogenic amylase method.
S Marcovina et al.
La Ricerca in clinica e in laboratorio, 14(3), 439-442 (1984-07-01)
[Amylase determination using p-nitrophenylmaltosides].
R Müller-Matthesius
Der Internist, 23(10), 575-578 (1982-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service