Skip to Content
Merck
All Photos(2)

Key Documents

N165

Sigma-Aldrich

Naloxone benzoylhydrazone

solid

Synonym(s):

[(5α)-4,5-Epoxy-3,14-dihydroxy-17-(2-propenyl)morphinan-6-ylidene]hydrazide benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27N3O4
CAS Number:
Molecular Weight:
445.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

yellow

solubility

DMSO: soluble
H2O: soluble (with the addition of 0.1 N HCl)
ethanol: soluble

storage temp.

−20°C

SMILES string

[H][C@@]12Oc3c(O)ccc4C[C@H]5N(CC[C@@]1(c34)[C@@]5(O)CC\C2=N/NC(=O)c6ccccc6)CC=C

InChI

1S/C26H27N3O4/c1-2-13-29-14-12-25-21-17-8-9-19(30)22(21)33-23(25)18(10-11-26(25,32)20(29)15-17)27-28-24(31)16-6-4-3-5-7-16/h2-9,20,23,30,32H,1,10-15H2,(H,28,31)/b27-18+/t20-,23+,25+,26-/m1/s1

InChI key

AKXCFAYOTIEFOH-XTNAHFASSA-N

Gene Information

Biochem/physiol Actions

Agonist for κ3 opioid receptors; antagonist for ORL1 and μ opioid receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronica Cox et al.
Neuropharmacology, 48(2), 228-235 (2005-02-08)
Naloxone benzoylhydrazone (NalBzoH) is a ligand used to study opioid receptors. It has been suggested to act at a novel kappa3 receptor but also appears to bind to classical opioid receptors, and possibly the ORL1 receptor. We have used opioid
Eve Salonius et al.
Journal of cellular physiology, 235(4), 3497-3507 (2019-09-26)
Cell therapy combined with biomaterial scaffolds is used to treat cartilage defects. We hypothesized that chondrogenic differentiation bone marrow-derived mesenchymal stem cells (BM-MSCs) in three-dimensional biomaterial scaffolds would initiate cartilaginous matrix deposition and prepare the construct for cartilage regeneration in
D Ichikawa et al.
Neuroreport, 12(8), 1757-1761 (2001-06-21)
J-113397 (1-[(3R,4R)-1-cyclooctylmethyl-3-hydroxymethyl-4-piperidyl]-3-ethyl-1,3-dihydro-2H-benzimidazol-2-one) is a recently developed antagonist of the opioid receptor-like 1 (ORL1) receptor. We compared the in vitro functional profile J-113397 on [35S]guanosine 5'-O-(gamma-thio)triphosphate (GTPgammaS) binding to mouse brain with that of [Phe1psi(CH2-NH)Gly2]nociceptin(1-13)NH2 and naloxone benzoylhydrazone (NalBzoH). J-113397 antagonized
Thomas M Tzschentke
Psychopharmacology, 172(1), 58-67 (2003-11-15)
Buprenorphine is widely used as an analgesic drug and it is also increasingly considered for maintenance and detoxification of heroin addicts. It is a potent micro -receptor partial agonist with a long duration of action. An inverted U-shaped dose-effect curve
D Paul et al.
The Journal of pharmacology and experimental therapeutics, 255(2), 769-774 (1990-11-01)
Naloxone benzoylhydrazone (NalBzoH) is a novel mixed agonist/antagonist. Against mu agonists, NalBzoH is a potent antagonist with a prolonged duration of action corresponding to its extremely slow rate of dissociation from mu receptors in binding assays. In the present studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service