Skip to Content
Merck
All Photos(4)

Key Documents

N1628

Sigma-Aldrich

Novobiocin sodium salt

≥90% (HPLC)

Synonym(s):

Albamycin, Albamycin Sodium, Cathomycin sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N2NaO11
CAS Number:
Molecular Weight:
634.61
Beilstein:
3892910
EC Number:
UNSPSC Code:
51286504
PubChem Substance ID:
NACRES:
NA.85

biological source

(STREPTOMYCES NIVEUS)

Assay

≥90% (HPLC)

form

powder

storage condition

(Tightly closed. Dry. )

color

white to yellow

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

[Na+].CO[C@@H]1[C@@H](OC(N)=O)[C@@H](O)[C@H](Oc2ccc3C([O-])=C(NC(=O)c4ccc(O)c(C\C=C(\C)C)c4)C(=O)Oc3c2C)OC1(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Novobiocin sodium salt is an antibiotic derived from soil bacteria. Novobiocin inhibits the action of bacterial DNA gyrase and belongs to a class of compounds called coumarin antibiotics. Coumarins are heterocyclic compounds with a wide range of biological activity, including antibacterial, anticoagulant, antioxidant, anti-inflammatory, antiviral, antitumor, and enzyme inhibitory effects.

Application

For the production of positively supercoiled plasmid DNA. Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. Inhibitor of retrovirus RNA-dependent DNA-polymerase.
Novobiocin Sodium Salt, an inhibitor of Hsp90, can be used:
  • in the research heat shock protein inhibition and to produce positively supercoiled plasmid DNA
  • in the research on the efficacy of probiotics
  • in the research on the efficacy of human monocytes and dendritic cells for T-cell replication

Biochem/physiol Actions

Mode of Action: Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II.
Antimicrobial spectrum: Gram-positive bacterial.
Novobiocin inhibits DNA synthesis by inhibiting the GyrB subunit of the Topoisomerase enzyme involved in energy transduction. Novobiocin acts as a competitive inhibitor of the ATPase reaction catalysed by GyrB.

Features and Benefits

High quality antibiotic suitable for mulitple research applications

Storage and Stability

Stored in frozen aliquots, Keep container tightly closed in a dry and well-ventilated place. Light sensitive

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of a Lactobacillus-based probiotic culture for the reduction of Salmonella enteritidis in neonatal broiler chicks
Higgins et al.
Poultry Science, 87, 27-31 (2008)
Relative efficacy of human monocytes and dendritic cells as accessory cells for T cell replication
Voorhis et al.
The Journal of Experimental Medicine, 58, 174-191 (1983)
Temporal effects of lactic acid bacteria probiotic culture on Salmonella in neonatal broilers
Higgins et al.
Poultry Science, Aug;86(8):, 1662-1666 (2007)
Yosef D Huberman et al.
Poultry science, 98(6), 2422-2431 (2019-01-29)
Salmonellosis caused by Salmonella Enteritidis is a widespread zoonosis and poultry products are an important source of infection. This study was carried out to evaluate the protection of different vaccination schedules in layers using a live commercial attenuated Salmonella Enteritidis
Margarita A Arreguin-Nava et al.
Animals : an open access journal from MDPI, 9(9) (2019-09-20)
The effect of Lactobacillus spp.-based probiotic candidates on Salmonella enterica serovar Enteritidis (SE) colonization was evaluated in two separate experiments. In each experiment, sixty-one day-of-hatch female turkey poults were obtained from a local hatchery. In both experiments, poults were challenged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service