MTCR1040
MTC™ Bio Fraction CollectorTube Rack
holds 80 x 1.5 /2.0 mL tubes, assorted colors, pack of 5 ea
Synonym(s):
tube holder, tube storage
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
Recommended Products
material
assorted colors
polypropylene
feature
holds 80 x 1.5 /2.0 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1040
Related Categories
General description
Compact, convenient 80 well racks designed to keep your tubes neatly and efficiently organized. Theses autoclavable polypropylene racks are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes.
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, assorted colors (B, G, R, Y, O), pack of 5
Features and Benefits
- 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
- Rainbow pack available — 5 colors
- Two frosted marking areas, on the bottom and side
- Raised numbers beside each well
- Recessed well bottoms for improved tube stability
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)