Skip to Content
Merck
All Photos(1)

Key Documents

MLS0001

Sigma-Aldrich

MISSION® LightSwitch Luciferase Assay Reagent

Fully optimized reporter system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.84

Quality Level

product line

MISSION®

storage temp.

−20°C

General description

The MISSION LightSwitch Luciferase Assay Reagent enables one-step reagent addition to measure luciferase reporter signal. The MISSION LightSwitch Luciferase Assay Reagent and the MISSION 3′UTR Lenti GoClone report constructs are a fully optimized reporter system that includes an improved reporter gene (RenSP), an optimized assay reagent, and a genome-wide collection of 3′UTR Lenti GoClone reporter constructs. The MISSION 3′UTR Lenti GoClone reporter constructs have been optimized for use with the MISSION LightSwitch Assay Reagent.

Application

MISSION® LightSwitch Luciferase Assay Reagent has been used to measure actin promoter activity.

Legal Information

LightSwitch is a trademark of Active Motif
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
SwitchGear Genomics is a trademark of SwitchGear Genomics

Kit Components Also Available Separately

Product No.
Description
SDS

  • MISSION® LightSwitch Assay Reagent Substrate

  • MISSION® LightSwitch Assay Reagent Buffer

  • MISSION® LightSwitch Assay Reagent Solvent

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janosch Hildebrand et al.
The Journal of investigative dermatology, 131(1), 20-29 (2010-09-10)
Here, we report a comprehensive investigation of changes in microRNA (miRNA) expression profiles on human keratinocyte (HK) differentiation in vitro and in vivo. We have monitored expression patterns of 377 miRNAs during calcium-induced differentiation of primary HKs, and have compared
Pieter Mestdagh et al.
Molecular cell, 40(5), 762-773 (2010-12-15)
The miR-17-92 microRNA cluster is often activated in cancer cells, but the identity of its targets remains elusive. Using SILAC and quantitative mass spectrometry, we examined the effects of activation of the miR-17-92 cluster on global protein expression in neuroblastoma
Federico Innocenti et al.
PLoS genetics, 7(5), e1002078-e1002078 (2011-06-04)
The discovery of expression quantitative trait loci ("eQTLs") can help to unravel genetic contributions to complex traits. We identified genetic determinants of human liver gene expression variation using two independent collections of primary tissue profiled with Agilent (n = 206) and Illumina
An NTD-associated polymorphism in the 3' UTR of MTHFD1L can affect disease risk by altering miRNA binding.
Minguzzi S
Human Mutation, 35(1), 96-104 (2014)
The PLA2R1-JAK2 pathway upregulates ERRa and its mitochondrial program to exert tumor-suppressive action.
Griveau A
Oncogene, 35(38), 5033-5042 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service