Skip to Content
Merck
All Photos(2)

Documents

MAK164

Sigma-Aldrich

Intracellular Hydrogen Peroxide Assay

sufficient for 200 fluorometric tests (green fluorescence)

Synonym(s):

Hydrogen Peroxide Detection in Cells

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 fluorometric tests (green fluorescence)

detection method

fluorometric

relevant disease(s)

aging/geriatric diseases; neurological disorders; pulmonary disorders; orthopedic diseases; cardiovascular diseases

storage temp.

−20°C

General description

Hydrogen peroxide, a reactive oxygen species produced through the metabolism of molecular oxygen, serves as both an intracellular signaling messenger and a source of oxidative stress. Hydrogen peroxide is generated in cells via multiple mechanisms such as the NOX-mediated ROS production by neutrophils and macrophages (respiratory burst) or by the dismutase of superoxide anions produced as a result of electron leak during mitochondrial respiration. Abnormal hydrogen peroxide production contributes to oxidative cell damage and the progression of diseases such as asthma, atherosclerosis, osteoporosis, and neurodegeneration.

Application

Intracellular hydrogen peroxide assay kit has been used to measure intracellular hydrogen peroxide levels.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

This kit is suitable for the detection of hydrogen peroxide in living cells and a variety of samples such as cell extracts and liquids.

Principle

The Intracellular Hydrogen Peroxide Assay Kit provides a simple and reproducible method to quantify hydrogen peroxide levels in living cells and in a variety of other samples such as cellular extracts. This kit utilizes a unique cell-permeable sensor that generates a fluorescent product (λex = 490/λem = 520 nm) after reaction with hydrogen peroxide that can be analyzed by a fluorescent microplate reader, flow cytometer, or fluorescent microscope.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongdong Wang et al.
iScience, 9, 14-26 (2018-10-28)
Therapeutic effects of photodynamic therapy (PDT) remain largely limited because of tumor hypoxia. Herein, we report safe and versatile nanocatalysts (NCs) for endogenous oxygen generation and imaging-guided enhanced PDT. The NCs (named as PSP) are prepared by coating Prussian blue
Maqusood Ahamed et al.
International journal of environmental research and public health, 17(21) (2020-11-12)
Combined exposure of single-walled carbon nanotubes (SWCNTs) and trace metal lead (Pb) in ambient air is unavoidable. Most of the previous studies on the toxicity of SWCNTs and Pb have been conducted individually. There is a scarcity of information on
Evidence of oxidative stress and mitochondrial dysfunction in spinocerebellar ataxia type 2 (SCA2) patient fibroblasts: effect of coenzyme Q10 supplementation on these parameters.
Cornelius N, et al.
Mitochondrion, 34, 103-114 (2017)
Maqusood Ahamed et al.
Journal of applied toxicology : JAT, 40(9), 1228-1238 (2020-03-29)
Broad application of reduced graphene oxide (rGO) and ubiquitous lead (Pb) pollution may increase the possibility of combined exposure of humans. Information on the combined effects of rGO and Pb in human cells is scarce. This work was designed to
Mohd Javed Akhtar et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-07)
This study aimed to generate a comparative data on biological response of yttrium oxide nanoparticles (Y2O3 NPs) with the antioxidant CeO2 NPs and pro-oxidant ZnO NPs. Sizes of Y2O3 NPs were found to be in the range of 35±10 nm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service