MAK132
Sulfate Assay Kit
sufficient for 200 tests (turbidimetric)
Synonym(s):
Sulfate Determination Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 200 tests (turbidimetric)
detection method
turbidimetric
relevant disease(s)
genitourinary disorders
storage temp.
room temp
General description
Inorganic sulfate is one of the most abundant anions in mammalian plasma. Sulfate plays important physiological roles in activating and detoxifying xenobiotics, steroids, neurotransmitters, and bile acids. Sulfate is needed for the biosynthesis of cerebroside sulfate, glycosaminoglycans, and heparin sulfate. Undersulfation of cartilage proteoglycans has been associated with human inherited osteochondro-dysplasia disorders. In mammals, sulfate homeostasis is regulated by the kidney. The majority of filtered sulfate is absorbed in the proximal tubules and only 5-20% of the filtered load is excreted into the urine.
Application
Sulfate Assay Kit has been used to determine the concentration of sulfate.
Features and Benefits
Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.
Suitability
Suitable for the detection of sulfate in biological fluids such as serum and urine.
Principle
The kit employs an improved method utilizes the quantitative formation of insoluble barium sulfate in polyethylene glycol. The turbidity measured at 600 nm is proportional to sulfate level in the sample.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Composition and structure of cell wall ulvans recovered from Ulva spp. along the Swedish west coast.
Carbohydrate polymers, 233, 115852-115852 (2020-02-16)
The cell wall polysaccharide ulvan was isolated from two species of the seaweed Ulva collected along the Swedish west coast. Acidic extraction was benchmarked against hot water extraction with enzymatic purification and against commercial ulvan. Extracted ulvan contained 11-18 %
Transcriptional regulation of the sodium-sulfate cotransporter NaSi-1 gene.
Cell Biochemistry and Biophysics, 36(2-3), 175-182 (2002)
Physiological Roles and Regulation of Mammalian Sulfate Transporters.
Physiological Reviews, 81(4) (2001)
Environmental microbiology, 23(7), 4017-4033 (2021-04-30)
Methoxylated aromatic compounds (MACs) are important components of lignin found in significant amounts in the subsurface. Recently, the methanogenic archaeon Methermicoccus shengliensis was shown to be able to use a variety of MACs during methoxydotrophic growth. After a molecular survey
Esters production via carboxylates from anaerobic paper mill wastewater treatment.
Bioresource Technology, 237 , 186-192 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service