Skip to Content
Merck
All Photos(2)

Key Documents

MAK109

Sigma-Aldrich

LPL Activity Assay Kit

Supplied by Roar Biomedical, Inc.

Synonym(s):

Lipoprotein lipase activity assay kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

gastrointestinal diseases; cardiovascular diseases

storage temp.

2-8°C

Gene Information

human ... LPL(4023)

General description

Lipoprotein lipase (LPL) hydrolyzes triglycerides associated with VLDL.

Application

LPL Activity Assay Kit may be used for detection of LPL activity in plasma samples of patients with hypertriglyceridemia.

Suitability

Suitable for the measurement of lipoprotein lipase activity in a variety of tissue samples

Principle

The LPL Activity Assay Kit includes a non-fluorescent substrate emulsion that becomes intensely fluorescent upon interaction with LPL and pre-hydrolyzed substrate for use as a standard to convert the fluorescence intensity reading to moles of reactant formed (λEx=370 nm/λEm=450 nm). The assay is not specific for LPL and will also detect hepatic lipase activity.

related product

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Szalowska et al.
Peptides, 32(5), 938-945 (2011-02-22)
GIP receptor knockout mice were shown to be protected from the development of obesity on a high fat diet, suggesting a role of GIP in the development of obesity. In our study we aimed to test the hypothesis if excess
Evemie Dubé et al.
Biology of reproduction, 87(1), 14-14 (2012-05-04)
Knowledge of the consequences of maternal obesity in human placental fatty acids (FA) transport and metabolism is limited. Animal studies suggest that placental uptake of maternal FA is altered by maternal overnutrition. We hypothesized that high maternal body mass index
Cedric H L Shackleton
Lipids, 47(1), 1-12 (2011-08-30)
In 1937 Butler and Marrian found large amounts of the steroid pregnanetriol in urine from a patient with the adrenogenital syndrome, a virilizing condition known to be caused by compromised adrenal secretion even in this pre-cortisol era. This introduced the
Tomomi Yamazaki et al.
Journal of lipid research, 53(10), 2024-2037 (2012-06-28)
Postprandial hyperlipidemia (lipemia) is a risk factor for atherosclerosis. However, mouse models of postprandial hyperlipidemia have not been reported. Here, we report that ddY mice display marked postprandial hypertriglyceridemia in response to dietary fat. In ddY mice, the fasting serum
Ismayil Tasdelen et al.
PloS one, 8(5), e64284-e64284 (2013-05-23)
The transcription factor PPARγ is the key regulator of adipocyte differentiation, function and maintenance, and the cellular target of the insulin-sensitizing thiazolidinediones. Identification and functional characterization of genes regulated by PPARγ will therefore lead to a better understanding of adipocyte

Protocols

Lipoprotein lipase (LPL) hydrolyzes triglycerides associated with VLDL.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service