Skip to Content
Merck
All Photos(1)

Key Documents

M9274

Sigma-Aldrich

Murashige and Skoog Basal Medium

suitable for plant cell culture, with sucrose and agar

Synonym(s):

MS Basal Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.72
Pricing and availability is not currently available.

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Murashige and Skoog Basal Medium has been used for culturing Arabidopsis transgenic seeds for cotyledon development,[1] pearl millet (Pennisetum glaucum),[2] and Rosmarinus ofcinalis L. for callus induction[3]

Formula variant

With the macro- and micronutrients, vitamins, sucrose and agar as described by Murashige and Skoog (1962).

Media Formulation

Quantity

Formulated to contain 42.4 grams of powder per liter of medium.

related product

Product No.
Description
Pricing

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baba Ngom et al.
Plant signaling & behavior, 12(9), e1356967-e1356967 (2017-08-02)
Cytosine DNA methylation is an epigenetic regulatory system used by plants to control gene expression. Methylation pattern always changes after abiotic stresses, pathogens and pest infections or after a treatment with salicylic acid (SA). The latter is a key player
Striking effects of melatonin on secondary metabolites produced by callus culture of rosemary (Rosmarinus officinalis L.)
Coskun Y, et al.
Plant Cell, Tissue and Organ Culture, 138(1), 89-95 (2019)
Antoine Fort et al.
The New phytologist, 209(2), 590-599 (2015-09-24)
Heterosis is the phenomenon whereby hybrid offspring of genetically divergent parents display superior characteristics compared with their parents. Although hybridity and polyploidy can influence heterosis in hybrid plants, the differential contributions of hybridity vs polyploidy to heterosis effects remain unknown.
Laure Audonnet et al.
Gene expression patterns : GEP, 25-26, 1-7 (2017-04-13)
Dimethylation of histone H3 lysine 9 (H3K9me2) is a heterochromatic mark linked to DNA methylation and gene repression. Removal of H3K9me2 from gene bodies by the jmjC histone demethylase IBM1/JMJ25 inhibits DNA methylation and derepresses gene expression. In this work
Smita P Chavan et al.
Applied microbiology and biotechnology, 89(6), 1701-1707 (2010-12-02)
The present study examined the effects of plant growth hormones, incubation period, biotic (Trametes versicolor, Mucor sp., Penicillium notatum, Rhizopus stolonifer, and Fusarium oxysporum) and abiotic (NaCl, MgSO(4), FeSO(4), ZnSO(4), and FeCl(3)) elicitors on cell growth and α-tocopherol and pigment

Questions

1–5 of 5 Questions  
  1. Would an incorrect pH level affect the setting of the agar?

    1 answer
    1. Yes, gelation is pH dependent. Adjusting the pH to 5.7 is recommended prior to heating to melt the agar and autoclaving to sterilize. Please see the link below to review the general plant culture medium preparation instructions available in the datasheet for product M5519 (formulation without agar):
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/361/537/m5519pis.pdf

      Helpful?

  2. How firm is the gel in the final sterile medium preparation of Murashige and Skoog Basal Medium, Product M9274?

    1 answer
    1. This formulation contains 8g/L of standard agar resulting in fairly firm and rigid culture medium.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What is the stability of the prepared Murashige and Skoog Basal Medium, Product M9274?

    1 answer
    1. This agar-containing medium can be stored for 3 to 6 months, depending on the stability of any additional supplements, if stored properly with no observable dehydration of the gel. If the medium appears to darken or pull away from the sides of the culture vessel, it should be discarded.

      Helpful?

  5. How is Murashige and Skoog Basal Medium, Product M9274 different from Product M5519?

    1 answer
    1. M9274 contains sucrose at 30 g/L and agar at 8 g/L; M5519 does not.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service