Skip to Content
Merck
All Photos(1)

Key Documents

M7898

Sigma-Aldrich

Monoclonal Anti-Thy 1.1 antibody produced in mouse

clone TN-26, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD90, Anti-CDw90

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TN-26, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

complement-mediated cytotoxicity assay: suitable
indirect immunofluorescence: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Thy1(21838)

General description

Monoclonal Anti-Thy 1.1 (mouse IgM isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Thymus cell antigen 1 (Thy 1.1) is a glycosylphosphatidyl-anchored protein. It belongs to the of the immunoglobulin superfamily. Thy 1.1 is expressed in stem cells, T-cells and neurons.

Specificity

Recognizes Thy 1.1 bearing cells and is less than 5% cross-reactive with Thy 1.2 bearing cells. Titer is 1:20,000 as determined by in vitro cytotoxicity against AKR/J thymocytes.

Immunogen

thymocytes from AKR/J (Thy 1.1) mice.

Application

Monoclonal Anti-Thy 1.1 antibody produced in mouse has been used in immunopanning and immunofluorescence.

Biochem/physiol Actions

Studies have shown that mice lacking the protein thymus cell antigen 1 (Thy 1.1) have an increased number of fibroblasts. It suppresses proliferation and enhances apoptosis in dermal fibroblasts, thereby modulating cell growth. This is carried out by regulating the action of β3 integrin.

Physical form

Solution in 0.1 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy A Neveu et al.
American journal of physiology. Cell physiology, 309(9), C616-C626 (2015-09-04)
Idiopathic pulmonary fibrosis is a progressive lung disease that increases in incidence with age. We identified a profibrotic lung phenotype in aging mice characterized by an increase in the number of fibroblasts lacking the expression of thymocyte differentiation antigen 1
Zhenghui Cheng et al.
BMC molecular and cell biology, 22(1), 21-21 (2021-04-09)
Schwann cells (SCs) play a crucial role in the repair of peripheral nerves. This is due to their ability to proliferate, migrate, and provide trophic support to axon regrowth. During peripheral nerve injury, SCs de-differentiate and reprogram to gain the
Glial-mediated neuroprotection: Evidence for the protective role of the NO-cGMP pathway via neuron-glial communication in the peripheral nervous system.
Thippeswamy T, et al.
Glia, 49(2), 197-210 (2005)
Yunsong Zhang et al.
Journal of molecular neuroscience : MN, 72(6), 1402-1412 (2022-05-17)
Cellular metabolism is essentially linked to tissue remodeling and organ regeneration. MOGS, a gene that encodes cellular metabolism-related protein mannosyl-oligosaccharide glucosidase, was found to be upregulated in nerve segments after peripheral nerve injury. Bioinformatic analyses identified upstream regulators of MOGS
Rapidly purified ganglion cells from neonatal mouse retinas allow studies of mitochondrial morphology and autophagy.
Zaninello M and Scorrano L
Pharmacological Research, 138, 16-24 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service