Skip to Content
Merck
All Photos(3)

Key Documents

M7753

Sigma-Aldrich

Maltoheptaose

≥60% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O36
CAS Number:
Molecular Weight:
1153.00
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

form

powder

concentration

≥60% (HPLC)

technique(s)

HPLC: suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)C(O)O[C@@H]7CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H72O36/c43-1-8-15(50)16(51)24(59)37(67-8)74-31-10(3-45)69-39(26(61)18(31)53)76-33-12(5-47)71-41(28(63)20(33)55)78-35-14(7-49)72-42(29(64)22(35)57)77-34-13(6-48)70-40(27(62)21(34)56)75-32-11(4-46)68-38(25(60)19(32)54)73-30-9(2-44)66-36(65)23(58)17(30)52/h8-65H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16+,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36?,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

BNABBHGYYMZMOA-QJBBZCPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maltoheptaose is a glucose heptamer.

Application

A partial hydrolysis of maltoheptaose has been used to prepare a dextran ladder in a study that assessed cationic labeling of oligosaccharides. The sensitivity of O-2-[aminoethyl]fluorescein was evaluated using maltoheptaose in a study that investigated the development of a novel fluorescent tag for the electrophoretic separation of oligosaccharides.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Partyka et al.
Journal of chromatography. A, 1267, 116-120 (2012-07-10)
Cationic derivatization of oligosaccharides by quaternary ammonium label was investigated for capillary electrophoretic separation with transient isotachophoretic preconcentration (t-ITP) as detected by capacitively coupled contactless conductivity detection (C4D). Dextran ladder, prepared by partial hydrolysis of dextran, isomaltotriose, maltopentaose and maltoheptaose
Svetlana E Soboleva et al.
Biomolecules, 9(6) (2019-05-31)
Lactoferrin (LF) is a Fe3+-binding glycoprotein, that was first recognized in milk and then in other epithelial secretions and barrier body fluids to which many different functions have been attributed to LF including protection from iron-induced lipid peroxidation, immunomodulation, cell
Nicole M Koropatkin et al.
Structure (London, England : 1993), 16(7), 1105-1115 (2008-07-10)
The human gut microbiota performs functions that are not encoded in our Homo sapiens genome, including the processing of otherwise undigestible dietary polysaccharides. Defining the structures of proteins involved in the import and degradation of specific glycans by saccharolytic bacteria
Artaches A Kazarian et al.
Analytica chimica acta, 662(2), 206-213 (2010-02-23)
This study describes the development of a novel fluorescent tag, O-2-[aminoethyl]fluorescein, for the separation of sugars by capillary electrophoresis with fluorescence detection using an argon ion laser. The tag was synthesised using three consecutive steps namely: esterification, alkylation and hydrolysis
Justyna M Dobruchowska et al.
Glycobiology, 22(4), 517-528 (2011-12-06)
Recently, a novel glucansucrase (GS)-like gene (gtfB) was isolated from the probiotic bacterium Lactobacillus reuteri 121 and expressed in Escherichia coli. The purified recombinant GTFB enzyme was characterized and turned out to be inactive with sucrose, the natural GS substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service