Skip to Content
Merck
All Photos(3)

Key Documents

M6824

Sigma-Aldrich

gamma-Mangostin

≥98% (HPLC)

Synonym(s):

1,3,6,7-Tetrahydroxy-2,8-bis(3,3-dimethylallyl)xanthone, Demethylmangostin, Normangostin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O6
CAS Number:
Molecular Weight:
396.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to yellow

solubility

methanol: 1 mg/mL, colorless to yellow

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC1=CC(OC2=CC(O)=C(CC=C(C)C)C(O)=C2C3=O)=C3C(CC=C(C)C)=C1O

InChI

1S/C23H24O6/c1-11(2)5-7-13-15(24)9-18-20(22(13)27)23(28)19-14(8-6-12(3)4)21(26)16(25)10-17(19)29-18/h5-6,9-10,24-27H,7-8H2,1-4H3

InChI key

VEZXFTKZUMARDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The fruit hulls of mangosteen contain three major bioactive compounds namely, α-, β-, and γ-mangostin.

Application

γ-Mangostin has been used to study its inhibitory effect on Transthyretin (TTR) fibrillization, a homotetrameric protein involved in human hereditary amyloidosis.

Biochem/physiol Actions

The xanthone derivatives, namely α-, β-, and γ-mangostin have various biological activities including antiproliferative and anti-inflammatory activity, enhancement of natural killer (NK) cell activity, and antiviral effect against human immunodeficiency virus (HIV). γ-Mangostin is also reported to inhibit cyclooxygenase (COX-2) activity and NO production, which in turn reduces the level of prostaglandin E2. It is found to induce cell death in human colon cancer cells.
Xanthone derivative isolated from the mangosteen hull (Garcinia mangostana). Exhibits antioxidant and anticancer properties. Mangosteen xanthones inhibit inflammation in human adipocytes which is important in understanding obesity-associated inflammation and the development of insulin resistance.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hui-Fang Chang et al.
Molecules (Basel, Switzerland), 15(12), 8953-8966 (2010-12-09)
Gliomas are a common type of primary brain tumor with glioblastoma multiforme accounting for the majority of human brain tumors. In this paper, high grade human malignant glioblastomas (MGs) including U87 MG and GBM 8401 were used to evaluate the
Monrudee Sukma et al.
Journal of ethnopharmacology, 135(2), 450-454 (2011-03-29)
γ-Mangostin is a xanthone found in the fruit hulls of Garcinia mangostana L., which have long been used in Southeast Asia as a traditional medicine for the treatment of abdominal pain, dysentery, wound infections, fever and convulsions. Recent studies have
Keigo Nakatani et al.
Molecular pharmacology, 66(3), 667-674 (2004-08-24)
We investigated the effect of gamma-mangostin purified from the fruit hull of the medicinal plant Garcinia mangostana on spontaneous prostaglandin E(2) (PGE(2)) genase release and inducible cyclooxy-2 (COX-2) gene expression in C6 rat glioma cells. An 18-h treatment with gamma-mangostin
N Chairungsrilerd et al.
Planta medica, 62(5), 471-472 (1996-10-01)
A crude methanolic extract of the fruit hull of Mangosteen, Garcinia mangostana L. inhibited the contractions of isolated thoracic rabbit aorta induced by histamine and serotonin. The extract of the fruit hull has been fractionated by silica gel chromatography, monitoring
Lih-Geeng Chen et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 688-693 (2007-11-22)
The fruit hull of Garcinia mangostana Linn (Guttiferae) is used as an anti-inflammatory drug in Southeast Asia. Two xanthones, alpha- and gamma-mangostins, were isolated from the fruit hull of G. mangostana, and both significantly inhibited nitric oxide (NO) and PGE(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service