Skip to Content
Merck
All Photos(1)

Key Documents

M6068

Sigma-Aldrich

Anti-phospho-Myosin Light Chain (pSer19) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MLC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human (predicted), chicken

technique(s)

western blot: 1:1,000 using chicken gizzard myosin II phosphorylated in vitro by PAK

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer19)

Gene Information

human ... MYL1(4632)

Related Categories

General description

Myosin light chains belongs to the calmodulin (CaM) gene family.
Rabbit polyclonal anti-phospho-Myosin Light Chain (pSer19) antibody detects chicken MLC. Results show that reactivity with the antibody requires phosphorylation of MLC by active kinase, and that only the peptide corresponding to MLC (pSer19) blocks the antibody signal, thereby demonstrating the specificity of the antibody. Human (100% homologous) has not been tested, but is expected to react.

Immunogen

synthetic phosphopeptide derived from the region of chicken MLC that contains serine 19 (serine 20 including the initiating methionine). Human (100% homologous) has not been tested, but is expected to cross-react.

Application

Anti-phospho-Myosin Light Chain (pSer19) antibody produced in rabbit has been used in the detection of phosphorylated myosin: in human lung fibroblasts and avian lens cells by immunoblotting by immunohistochemical detection in rabbit aorta samplesby immunofluorescence detection in human submandibular gland tissues

Biochem/physiol Actions

Myosin light chain is a subunit of the conventional myosins, e.g., myosin II. In smooth muscle and nonmuscle cells, conventional myosins mediate a wide variety of contractile events including cytokinesis, cell motility, and smooth muscle contraction. MLC is phosphorylated by multiple serine-threonine kinases such as Rho-kinase and PAK, however, myosin light chain kinase (MLCK) acts as the primary kinase. Contractile activity of conventional myosins is regulated by phosphorylation of MLC on several residues. Phosphorylation of serine 19, the major phosphorylation site and the preferred site for MLCK, activates myosin motor activity under physiological conditions.
Myosin light chains functions to maintain integrity of myosin protein. Mutations in the myosin light chain is implicated in familial hypertrophic cardiomyopathy.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (± 0.1), 50% glycerol with 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of actomyosin disruptors on the mechanical integrity of the avian crystalline lens
Won GJ et al.
Molecular Vision, 21(9), 98-98 (2015)
Shuichi Asano et al.
Physiological reports, 5(9) (2017-05-17)
In patients with pulmonary diseases such as idiopathic pulmonary fibrosis and severe acute respiratory distress syndrome, progressive pulmonary fibrosis is caused by dysregulated wound healing via activation of fibroblasts after lung inflammation or severe damage. Migration of fibroblasts toward the
Matrix stiffness regulates migration of human lung fibroblasts
Asano S, et al.
Physiological Reports, 5(9), e13281-e13281 (2017)
Myosin essential light chain in health and disease
Hernandez OM, et al.
American Journal of Physiology. Heart and Circulatory Physiology, 292(4), H1643-H1654 (2007)
Myosin light chain kinase inhibitor ML7 improves vascular endothelial dysfunction via tight junction regulation in a rabbit model of atherosclerosis
Cheng X, et al.
Molecular Medicine Reports, 12(3), 4109-4116 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service