Skip to Content
Merck
All Photos(1)

Key Documents

M3682

Sigma-Aldrich

Anti-MAP Kinase, Monophosphorylated Tyrosine antibody ,Mouse monoclonal

clone ERK-PY193, purified from hybridoma cell culture

Synonym(s):

Anti-pY-ERK

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

ERK-PY193, monoclonal

form

buffered aqueous solution

mol wt

antigen, ERK-1 44 kDa
antigen, ERK-2 42 kDa

species reactivity

rat, human

concentration

~2 mg/mL

technique(s)

capture ELISA: suitable
immunocytochemistry: suitable
microarray: suitable
western blot: 1-5 μg/mL using cell extract of rat fibroblasts cell line, Rat1, activated with sorbitol

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr)

Gene Information

General description

Monoclonal Anti-MAP Kinase, Mono-phosphorylated Tyrosine (pY-ERK) (mouse IgG1 isotype) is derived from the ERK-PY193 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide sequence corresponding to the phosphorylated form of ERK-activation loop, conjugated to KLH. The mitogen-activated protein kinase (MAPK) superfamily of enzymes is involved in many signaling pathways. This family includes the ERK1/2 (extracellular signal-regulated protein kinase, also termed p42/p44 MAPK), JNK (c-Jun N-terminal protein kinase, also termed stress-activated protein kinase, SAPK1), and p38 MAPK (also termed SAPK2) subfamilies.

Specificity

The antibody reacts with the monophosphorylated tyrosine form of MAP kinases (ERK1 and ERK2). It does not recognize the non-phosphorylated, diphosphorylated, and the monophosphorylated threonine forms of ERK/MAP kinases, or the diphosphorylated forms of JNK and p38 MAPK. The epitope recognized by the antibody contains the phosphorylated tyrosine residue within the regulatory site of MAP kinase (e.g.,Tyr185 in ERK-2). Cross-reactivity has been observed with the monophosphorylated tyrosine peptide of JNK.

Immunogen

synthetic peptide HTGFLTEpYVAT, corresponding to the phosphorylated form of ERK-activation loop.

Application

Monoclonal Anti-MAP Kinase, Monophosphorylated Tyrosine antibody has been used in:
  • immunoblotting
  • enzyme linked immunosorbent assay (ELISA)
  • dot-blot
  • immunocytochemistry
  • western blotting
  • immunofluorescence

Biochem/physiol Actions

MAPK kinase (MAPKK) is the immediate upstream activator of the MAPK, MAPKK kinase (MAP3K), and MAP3K kinase (MAP4K) for the enzymes further upstream, respectively. The kinases in the MAPK level are activated by phosphorylation of both tyrosine (Y) and threonine (T) residues organized in a TXY motif. The residue in between the two phosphorylated residues determines the specificity of activation of the MAPKs. Phosphorylation of both tyrosine and threonine is essential for the full activation of all MAPKs.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Prepared from a culture supernatant of bioreactor grown hybridoma.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudhakaran Prabakaran et al.
Biophysical journal, 106(12), 2720-2728 (2014-06-19)
Mathematical models are extensively employed to understand physicochemical processes in biological systems. In the absence of detailed mechanistic knowledge, models are often based on network inference methods, which in turn rely upon perturbations to nodes by biochemical means. We have
Mammalian MAP kinase signalling cascades
Chang L and Karin M
Nature, 410(6824), 37-37 (2001)
Paradoxical results in perturbation-based signaling network reconstruction
Prabakaran S, et al.
Biophysical Journal, 106(12), 2720-2728 (2014)
Monophosphothreonyl extracellular signal-regulated kinases 1 and 2 (ERK1/2) are formed endogenously in intact cardiac myocytes and are enzymically active.
Sugden PH, et al.
Cellular Signalling, 23(2), 468-477 (2011)
H Cha et al.
The Journal of cell biology, 153(7), 1355-1367 (2001-06-27)
Phosphorylation of the extracellular signal-regulated kinases (ERKs) on tyrosine and threonine residues within the TEY tripeptide motif induces ERK activation and targeting of substrates. Although it is recognized that phosphorylation of both residues is required for ERK activation, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service