Skip to Content
Merck
All Photos(1)

Key Documents

M2521

Sigma-Aldrich

IgM, Lambda from murine myeloma

clone MOPC 104E, ascites fluid, lyophilized powder

Synonym(s):

Mouse IgM-λ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

clone

MOPC 104E, monoclonal

form

lyophilized powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The MOPC-104E tumor line that produces mouse IgM is a mineral oil induced plasmacytoma originated and carried intraperitoneally in BALB/c mice. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE.[1] IgM is present on the surface of B lymphocytes in its monomeric form.[2]

Specificity

Antigen Specificity: α(1→3) glucose

Application

IgM, λ from murine myeloma has been used as control in chromatin immunoprecipitation analysis.[3] It has also been used in immunohistochemitsry.[4]
IgM, λ from murine myeloma was used as a control antibody in ChIP analysis of GATA-1-null and GATA-1-positive G1E cells.[5]

Biochem/physiol Actions

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.[6]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GATA-1-mediated Transcriptional Repression Yields Persistent Transcription Factor IIB-Chromatin Complexes
Martowicz ML, et al.
The Journal of biological chemistry, 281(49), 37345-37352 (2006)
D Chen et al.
The American journal of physiology, 277(3), H1100-H1106 (1999-09-14)
Abnormal proliferation of vascular smooth muscle cells (VSMCs) is thought to play an important role in the pathogenesis of atherosclerosis and restenosis. Previous studies have implicated the extracellular matrix protein thrombospondin-1 (TSP1) in mitogen-dependent proliferation of VSMCs. In this study
Melissa L Martowicz et al.
The Journal of biological chemistry, 281(49), 37345-37352 (2006-09-12)
The hematopoietic GATA factors GATA-1 and GATA-2, which have distinct and overlapping roles to regulate blood cell development, are reciprocally expressed during erythropoiesis. GATA-1 directly represses Gata2 transcription, and reduced GATA-2 synthesis promotes red blood cell development. Gata2 repression involves
Robert McGlynn et al.
The Journal of comparative neurology, 480(4), 415-426 (2004-11-24)
The mucopolysaccharidoses (MPSs) are a complex family of lysosomal storage disorders characterized by failure to degrade heparan sulfate (HS) and/or other types of glycosaminoglycans (GAGs) secondary to the absence of specific lysosomal enzymes. An accompanying storage of glycosphingolipids (GSLs), most
The Immunoglobulins: Structure and Function (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service