Skip to Content
Merck
All Photos(2)

Key Documents

M0388

Sigma-Aldrich

6α-Methyl-11β-hydroxyprogesterone

Synonym(s):

11β-Hydroxy-6α-methyl-4-pregnene-3,20-dione, Medrysone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

form

solid

shipped in

ambient

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@]4(C)[C@H](CC[C@@]24[H])C(C)=O

InChI

1S/C22H32O3/c1-12-9-15-17-6-5-16(13(2)23)22(17,4)11-19(25)20(15)21(3)8-7-14(24)10-18(12)21/h10,12,15-17,19-20,25H,5-9,11H2,1-4H3/t12-,15-,16+,17-,19-,20+,21-,22+/m0/s1

InChI key

GZENKSODFLBBHQ-ILSZZQPISA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Management of vernal conjunctivitis with steroid induced glaucoma--a comparative study.
S Mithal et al.
Indian journal of ophthalmology, 35(5-6), 298-301 (1987-01-01)
N L Verbey et al.
Current eye research, 7(6), 557-561 (1988-06-01)
The effect of topical administration of 3-isobutyl-methyl-xanthine (IBMX), a potent phosphodiesterase inhibitor, was studied on an experimentally provoked uveitis in rabbits. After presensitization with an intravitreal injection of human serum albumin (HSA), intravenous antigenic challenge induces blood-aqueous barrier breakdown and
J F Recasens et al.
Ophthalmic research, 22(1), 12-18 (1990-01-01)
Superoxide dismutase activity (SOD) was measured in the irides of control animals and 24 h after the intravitreal administration of Escherichia coli endotoxin. A 3-fold increase in SOD was noted (p less than 0.001). To protect against the inflammatory process
K Green et al.
Current eye research, 8(3), 317-320 (1989-03-01)
The interaction between a topically administered progesterone, medrysone, and a steroid antagonist, mifepristone, on rabbit intraocular pressure (IOP) has been determined. Medrysone alone increased IOP significantly above parallel controls over the first three weeks; this increase was not sustained. When
J S Mindel et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 98(9), 1577-1578 (1980-09-01)
In age-, race-, and sex-matched groups, the relative abilities of medrysone, fluorometholone, and dexamethasone phosphate to elevate intraocular pressure were compared in a masked manner. Compliance was controlled by drops being given by nursing personnel. The 18 male hospitalized volunteers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service