Skip to Content
Merck
All Photos(1)

Key Documents

M0195

Sigma-Aldrich

MAPKAPK2 human

recombinant, expressed in HEK 293 cells, ≥60 units/mg protein, buffered aqueous glycerol solution

Synonym(s):

MAP kinase activated protein kinase 2, MAPKAP Kinase 2 human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in HEK 293 cells

Quality Level

form

buffered aqueous glycerol solution

specific activity

≥60 units/mg protein

mol wt

45-60 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPKAPK2(9261)

Biochem/physiol Actions

A serine/threonine protein kinase activated by the p38 MAP kinase in response to various signals including mitogenic and stress stimuli. The physiological substrate of MAPKAPK2
MAPKAPK2 is a serine/threonine protein kinase activated by the p38 MAP kinase in response to various signals including mitogenic and stress stimuli. MAPKAPK2 is distinguished from the RSK family of ribosomal protein S6 kinases (also termed MAPKAPK1), which are activated by the MAP kinases, ERK1, and ERK2 based on its substrate specificity and amino acid sequence. Upon activation, p38 MAP kinase activates the downstream MAPKAPK2 by phosphorylation of the threonine residue in the Pro-Thr motif in response to heat shock or mitogenic stimuli. Activated MAPKAPK2 phosphorylates the small heat shock protein HSP25/HSP27, both in vitro and in vivo, at serine residues. Thus, HSP25/HSP27 seems to be the physiological substrate of MAPKAPK2.

Unit Definition

One unit will incorporate 1.0 nmole of phosphate into HSP 25 per minute at 30 °C at pH 7.4.

Physical form

Solution in 50 mM Tris-HCl, pH 7.4, 100 mM NaCl, 0.05 mM EDTA, 2 mM DTT, and 10% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung-Jen Wei et al.
Cell death & disease, 11(5), 368-368 (2020-05-16)
Despite the improvement in clinical outcome with 13-cis-retinoic acid (13-cisRA) + anti-GD2 antibody + cytokine immunotherapy given in first response ~40% of high-risk neuroblastoma patients die of recurrent disease. MYCN genomic amplification is a biomarker of aggressive tumors in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service