L8775
Lectin from Galanthus nivalis (snowdrop)
agarose conjugate, saline suspension
Synonym(s):
Galanthus nivalis lectin, GNL
About This Item
Recommended Products
conjugate
agarose conjugate
Quality Level
form
saline suspension
extent of labeling
2-4 mg protein per mL
matrix
cross-linked 4% beaded agarose
capacity
5-10 mg/mL binding capacity (yeast mannan)(pH7.2 at 25 °C)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | L8275 | L1882 | U5632 |
---|---|---|---|
2-8°C | 2-8°C | 2-8°C | 2-8°C |
agarose conjugate | - | agarose conjugate | - |
cross-linked 4% beaded agarose | - | cross-linked 4% beaded agarose | Fast flow highly cross-linked 4% beaded agarose |
200 | 200 | 200 | 200 |
2-4 mg protein per mL | - | 5-10 mg per mL | 7-15 mg per mL |
Biochem/physiol Actions
Physical form
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service