L8376
Liver acetone powder from pigeon
Synonym(s):
Liver powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.25
Recommended Products
Application
Liver acetone powder from pigeon has been used in the estimation of pantothenic acid in food samples. It has also been used in the preparation of pigeon liver amidase for the digestion of pantothenate compounds in rat liver and blood samples.
Preparation Note
Contains NAD kinase activity.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tsutomu Fukuwatari et al.
Journal of nutritional science and vitaminology, 56(1), 19-26 (2010-04-01)
Recent studies showed that the urinary excretion of the water-soluble vitamins can be useful as a nutritional index. To determine how fasting affects urinary excretion of water-soluble vitamins, a human study and an animal experiment were conducted. In the human
C Pakin et al.
Journal of chromatography. A, 1035(1), 87-95 (2004-05-01)
A method to determine the content of free pantothenic acid in various foods by reverse phase liquid chromatography-fluorimetry is reported. It includes a purification of the samples by successive passages through anion and cation exchange cartridges and a post-column derivatization
Pierre Andrieux et al.
Journal of AOAC International, 95(1), 143-148 (2012-04-04)
A novel method was developed and single-laboratory validated for the determination of free pantothenic acid (vitamin B5) in a wide range of infant and adult fortified food products. The method combines simple sample preparation and chromatographic analysis using ultra-performance LC
Nima N Naseri et al.
Journal of neuropathology and experimental neurology, 74(6), 527-537 (2015-05-16)
Glucose metabolism is reduced in the brains of patients with Huntington disease (HD). The mechanisms underlying this deficit, its link to the pathology of the disease, and the vulnerability of the striatum in HD remain unknown. Abnormalities in some of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service