Skip to Content
Merck
All Photos(1)

Key Documents

L8008

Sigma-Aldrich

Luteinizing hormone releasing hormone

≥97% (HPLC)

Synonym(s):

GnRH, Gonadotropin-releasing hormone, LH-RH, [Gly-OH10]-LH-RH, pGlu-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H74N16O14
CAS Number:
Molecular Weight:
1183.27
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Luteinizing hormone releasing hormone, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)CNC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CO)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(Cc4cnc[nH]4)NC(=O)C5CCC(=O)N5)C(=O)NC(CCCNC(N)=N)C(=O)N6CCCC6C(=O)NCC(O)=O

InChI

1S/C55H74N16O14/c1-29(2)19-38(49(80)66-37(9-5-17-59-55(56)57)54(85)71-18-6-10-43(71)53(84)62-26-46(76)77)65-45(75)25-61-47(78)39(20-30-11-13-33(73)14-12-30)67-52(83)42(27-72)70-50(81)40(21-31-23-60-35-8-4-3-7-34(31)35)68-51(82)41(22-32-24-58-28-63-32)69-48(79)36-15-16-44(74)64-36/h3-4,7-8,11-14,23-24,28-29,36-43,60,72-73H,5-6,9-10,15-22,25-27H2,1-2H3,(H,58,63)(H,61,78)(H,62,84)(H,64,74)(H,65,75)(H,66,80)(H,67,83)(H,68,82)(H,69,79)(H,70,81)(H,76,77)(H4,56,57,59)

InChI key

IGACZQNUZMBGFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly

Biochem/physiol Actions

Luteinizing hormone releasing hormone (LH-RH) is used to release follicle-stimulating hormone (FSH) and luteinizing hormone (LH) from the anterior pituitary and to study many normal and pathological processes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Mathew et al.
Biology of reproduction, 100(2), 365-380 (2018-09-12)
This study investigated bovine conceptus-induced modifications to the endometrial transcriptome related to effects of interferon tau (IFNT), conceptus origin (in vivo vs. in vitro), and conceptus sex. In vitro (IVF) or in vivo (superovulation and artificial insemination, AI) produced blastocysts
Mohannad Alharbi et al.
Asian journal of andrology, 22(4), 348-353 (2019-10-02)
Sperm DNA fragmentation (SDF) has been linked with male infertility, and previous studies suggest that SDF can have negative influence on pregnancy outcomes with assisted reproduction. We performed a retrospective review of consecutive couples with a high SDF level that
Sandro C Esteves et al.
Frontiers in endocrinology, 10, 917-917 (2020-02-11)
This multicenter study evaluated the reliability of the recently published ART calculator for predicting the minimum number of metaphase II (MII) oocytes (MIImin) to obtain at least one euploid blastocyst in patients undergoing in vitro fertilization/intracytoplasmic sperm injection (IVF/ICSI). We
Runa Özelçi et al.
Turkish journal of medical sciences, 49(4), 1138-1144 (2019-07-12)
Diminished ovarian reserve (DOR) represents a major challenge in reproductive medicine, as it is often associated with poor ovarian stimulation response, high cycle cancellation rate, and low pregnancy rate. The aim of the present study is to compare the
Constantine A Simintiras et al.
Biology of reproduction, 100(3), 672-685 (2018-11-06)
Pregnancy establishment in cattle is contingent on conceptus elongation-a fundamental developmental event coinciding with the time during which most pregnancies fail. Elongation in vivo is directly driven by uterine secretions, indirectly influenced by systemic progesterone concentrations, and has yet to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service