Skip to Content
Merck
All Photos(1)

Key Documents

L2018

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli K-235

purified by gel-filtration chromatography

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (K-235)

Quality Level

form

lyophilized powder

purified by

gel-filtration chromatography

impurities

<1% Protein

color

white to yellow cast

solubility

water: 4.90-5.10 mg/mL, cloudy to hazy, colorless to faintly yellow

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is extracted from E. coli serotype K235 and purified by gel filtration. The source strain is ATCC 13027.

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Waleed Al Abdulmonem et al.
Journal of reproductive immunology, 141, 103152-103152 (2020-06-11)
During first trimester of human pregnancy, the maternal system develops immunity against infection and to provide protection of allogeneic foetus from abortion. This study was undertaken to determine the role of trophoblast specific CD74 isoforms in first trimester trophoblast derived
Peipei Zhang et al.
Science advances, 7(43), eabi9471-eabi9471 (2021-10-23)
[Figure: see text].
Okito Hashimoto et al.
Brain, behavior, and immunity, 90, 319-331 (2020-09-21)
Allergic dermatitis (AD), associated with pruritus and itchiness, is one of the major stressful conditions early in life. AD also influences the incidence of neuropsychiatric disorders and developmental disorders through neuro-immune interactions. To the best of our knowledge, there is
Jonathan J Hansen et al.
Inflammatory bowel diseases, 15(6), 890-899 (2009-01-13)
While others have described gene expression patterns in humans with inflammatory bowel diseases and animals with chemically induced colitis, a genome-wide comparison of gene expression in genetically susceptible animals that develop spontaneous colitis has not been reported. We used microarray
Tao Ni et al.
Science advances, 6(5), eaax8286-eaax8286 (2020-02-18)
Perforin-2 (MPEG1) is thought to enable the killing of invading microbes engulfed by macrophages and other phagocytes, forming pores in their membranes. Loss of perforin-2 renders individual phagocytes and whole organisms significantly more susceptible to bacterial pathogens. Here, we reveal

Articles

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Related Content

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Lipopolysaccharide (LPS) is a major component of Gram-negative bacteria, important for microbiological research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service