Skip to Content
Merck
All Photos(4)

Key Documents

L0259

Sigma-Aldrich

Lucifer Yellow CH dilithium salt

Powder

Synonym(s):

6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulfonic acid dilithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Li2N5O9S2
CAS Number:
Molecular Weight:
457.25
Beilstein:
4644601
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Lucifer Yellow CH dilithium salt, fluorescent stain

form

powder

color

faint orange to very dark orange

solubility

H2O: 1 mg/mL

fluorescence

λex 428 nm; λem 540 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Li+].[Li+].NNC(=O)NN1C(=O)c2cc(cc3c(N)c(cc(C1=O)c23)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C13H11N5O9S2.2Li/c14-10-5-1-4(28(22,23)24)2-6-9(5)7(3-8(10)29(25,26)27)12(20)18(11(6)19)17-13(21)16-15;;/h1-3H,14-15H2,(H2,16,17,21)(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2

InChI key

RPKCZJYDUKVMGF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Lucifer Yellow CH is the hydrazine (-NH-CO-NH-NH2) form of the dye compared to Lucifer Yellow VS, which is the vinyl sulfone (-Ph-SO2-CH=CH2) form. Lucifer Yellow CH dilithium salt is a fluorescent dye(1) comprising of amino, sulfo and phenyl-N-hydrazinocarbonylamino substituents on the naphthalimide chromophore. The hydrophilic nature of Lucifer yellow CH underlies its numerous applications as a stain of living cells and tissues.

Application

A highly fluorescent dye, useful in marking nerve cells.
Lucifer yellow CH has been used to stain nerve cells and to demonstrate electrical coupling between hamster oocytes and cumulous cells during meiotic maturation in vivo and in vitro.
Lucifer Yellow CH is mainly used as a tracer dye and useful in marking nerve cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic, fluorescent dye and electrical coupling between hamster oocytes and cumulus cells during meiotic maturation in vivo and in vitro.
Racowsky, C. and Satterlie, R.A.
Developmental Biology, 108, 191-202 (1985)
Mariane C Vicente et al.
Experimental neurology, 328, 113250-113250 (2020-02-24)
The locus coeruleus (LC) is a pontine nucleus important for respiratory control and central chemoreception. It is affected in Alzheimer's disease (AD) and alteration of LC cell function may account for respiratory problems observed in AD patients. In the current
Qi-You Li et al.
Experimental eye research, 202, 108305-108305 (2020-10-21)
The biosafety and efficiency of transplanting retinal pigment epithelial (RPE) cells derived from both human embryonic stem cells (hESCs) and induced pluripotent stem cells (iPSCs) have been evaluated in phase I and phase II clinical trials. For further large-scale application
Functional connections between cells as revealed by dye-coupling with a highly fluorescent naphthalimide tracer.
W W Stewart
Cell, 14(3), 741-759 (1978-07-01)
Elisabet Miro-Casas et al.
Cardiovascular research, 83(4), 747-756 (2009-05-23)
Connexin43 is present at the inner membrane of cardiomyocyte mitochondria (mCx43), but its function remains unknown. In this study we verified the presence of mCx43 by a mass spectrometry-based proteomic approach in purified mitochondrial preparations from mouse myocardium and determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service