Skip to Content
Merck
All Photos(4)

Key Documents

K3269

Sigma-Aldrich

Anti-Kindlin-2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FERMT2, Anti-Fermitin family homolog 2, Anti-KIND2, Anti-MIG2, Anti-Mitogen inducible gene 2 protein, Anti-PLEKHC1, Anti-Pleckstrin homology domain containing, family C member 1, Anti-UNC112, Anti-UNC112B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~72 kDa

species reactivity

mouse, rat, canine, human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2-5 μg/mL using canine MDCK cells
western blot: 1-2 μg/mL using whole extracts of human HeLa, mouse P19 or rat AT3B-1 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Anti-kindlin-2 recognizes human, mouse, rat and canine kindlin-2. Kindlin-2, also known as fermitin family member 2 (FERMT2) or pleckstrin homology domain-containing family C member 1 (PLEKHC1), is encoded by the gene mapped to human chromosome 14q22.1. Kindlin-2 is a member of the kindlin protein family and has a ubiquitous expression pattern.

Application

Anti-kindlin-2 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Kindlin-2 acts as a focal adhesion protein and controls breast cancer progression by promoting genome instability. Kindlin-2, as an integrin-interacting protein, plays a vital role in integrin activation and integrin-mediated cell-extracellular matrix (ECM) interaction. Kindlin-2 expression is associated with poor prognosis of patients with hepatocellular carcinoma (HCC) and pancreatic ductal adenocarcinomas (PDACs). Therefore, kindlin-2 can be considered as a potential biomarker for patients with HCC and PDACs.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kindlin-2 siRNA inhibits vascular smooth muscle cell proliferation, migration and intimal hyperplasia via Wnt signaling
Wu X, et al.
International Journal of Molecular Medicine, 37, 436-444 (2016)
Bing Li et al.
Science China. Life sciences, 62(2), 225-234 (2018-11-22)
Kindlin-2, an integrin-interacting protein, regulates breast cancer progression. However, currently, no animal model to study the role of Kindlin-2 in the carcinogenesis of mammary gland is available. We established a Kindlin-2 transgenic mouse model using a mammary gland-specific promoter, mammary
Xiaowei Xue et al.
Cancer medicine, 9(14), 4971-4979 (2020-05-22)
Breast cancer was the most frequent and the second most deadly cancer in women in 2018 in China; thus, early diagnosis of breast cancer is important. Studies have reported that tissue stiffness promotes cancer progression through increased collagen or fibrosis.
Si-Yu Guan et al.
FEBS letters, 592(1), 112-121 (2017-12-14)
Focal adhesion (FA) proteins, kindlin-2 and integrin-linked kinase (ILK), regulate cell adhesion and migration. ILK interacts with and promotes kindlin-2 targeting to FAs. Leu353 and Leu357 in kindlin-2 have been reported to be important for the interaction between kindlin-2 and
Qiu-Yue Zhang et al.
Oncotarget, 7(26), 39044-39050 (2016-10-27)
A recent meta-analysis of genome-wide association studies (GWAS) in population of Caucasian identified a single nucleotide polymorphism (SNP) rs17125944 in the FERMT2 gene as a new susceptibility locus for late-onset Alzheimer's disease (LOAD). In order to validate the association of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service