Skip to Content
Merck
All Photos(1)

Key Documents

I9759

Sigma-Aldrich

Ile-Pro-Ile

≥97% (HPLC)

Synonym(s):

Diprotin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H31N3O4
CAS Number:
Molecular Weight:
341.45
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCC(C)C(N)C(=O)N1CCCC1C(=O)NC(C(C)CC)C(O)=O

InChI

1S/C17H31N3O4/c1-5-10(3)13(18)16(22)20-9-7-8-12(20)15(21)19-14(17(23)24)11(4)6-2/h10-14H,5-9,18H2,1-4H3,(H,19,21)(H,23,24)

InChI key

JNTMAZFVYNDPLB-UHFFFAOYSA-N

Amino Acid Sequence

Ile-Pro-Ile

Application

Ile-Pro-Ile has been used as a supplement for blood collection for hormone measurement and used in Islets cell culture to stabilize glucagon-like peptide-1 (GLP-1). It has also been used to inhibit the breakdown of gonadotropin-inhibitory hormone (GnIH) following serum collection.

Biochem/physiol Actions

Dipeptidyl peptidase IV Inhibitor. Inhibits entry of HIV-1 or HIV-2
Inhibitor of dipeptidyl peptidase IV. Inhibits entry of HIV-1 or HIV-2 into T lymphoblastoid and monocytoid cell lines.

Features and Benefits

This compound is featured on the Neuropeptidases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of gonadotropin-inhibitory hormone on luteinizing hormone secretion in humans
George JT, et al.
Clinical Endocrinology, 86(5), 731-738 (2017)
F Samuel van Nierop et al.
Clinical nutrition (Edinburgh, Scotland), 36(6), 1615-1620 (2016-10-25)
Bile acids (BA) are pleiotropic hormones affecting glucose and lipid metabolism. The physiochemical properties of different BA species affect their enterohepatic dynamics and their affinity for bile acid receptors. The BA pool composition is altered in patients with type 2
Gemma Pujadas et al.
Molecular metabolism, 39, 101008-101008 (2020-05-12)
Glucose-dependent insulinotropic polypeptide (GIP) conveys information from ingested nutrients to peripheral tissues, signaling energy availability. The GIP Receptor (GIPR) is also expressed in the bone marrow, notably in cells of the myeloid lineage. However, the importance of gain and loss
Pancreatic alpha cell-derived glucagon-related peptides are required for beta cell adaptation and glucose homeostasis
Traub S, et al.
Cell Reports, 18(13), 3192-3203 (2017)
Carlos García-Aparicio et al.
Journal of medicinal chemistry, 49(17), 5339-5351 (2006-08-18)
Here we describe a novel type of enzyme-based prodrug approach in which a dipeptide moiety is linked to a nonpeptidic therapeutic drug through an amide bond which is specifically cleaved by the dipeptidyl-peptidase IV (DPP IV/CD26) enzyme activity present in

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service