Skip to Content
Merck
All Photos(1)

Key Documents

I9532

Sigma-Aldrich

Icilin

≥97% (HPLC), solid

Synonym(s):

1-(2-Hydroxyphenyl)-4-(3-nitrophenyl)-1,2,3,6-tetra­hydro­pyrimidin-2-one, AG-3-5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N3O4
CAS Number:
Molecular Weight:
311.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

color

yellow

solubility

DMSO: 15 mg/mL

SMILES string

Oc1ccccc1N2CC=C(NC2=O)c3cccc(c3)[N+]([O-])=O

InChI

1S/C16H13N3O4/c20-15-7-2-1-6-14(15)18-9-8-13(17-16(18)21)11-4-3-5-12(10-11)19(22)23/h1-8,10,20H,9H2,(H,17,21)

InChI key

RCEFMOGVOYEGJN-UHFFFAOYSA-N

General description

Icilin is a synthetic cooling agent and modulates human transient receptor potential cation channel (TRPA1). Icilin inhibits transient receptor potential cation channel subfamily M member 8 (TRPM8) and modulates calcium release. Icilin also suppresses transient receptor potential vanilloid subtype 3 (TRPV3).

Application

Icilin has been used as cooling agent in anterior ventromedial preoptic area (VMPO) neurons.

Biochem/physiol Actions

Icilin is a potent agonist at the CMR1 cold- and menthol-sensitive receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Warm-sensitive neurons that control body temperature
Tan CL, et al.
Cell, 167(1), 47-59 (2016)
Supercooling agent icilin blocks a warmth-sensing ion channel TRPV3
Sherkheli MA, et al.
TheScientificWorldJournal, 2012(6), 859-869 (2012)
Inhibition of TRPM8 by icilin distinct from desensitization induced by menthol and menthol derivatives
Kuhn FJP, et al.
The Journal of Biological Chemistry, 284(7), 4102-4111 (2009)
Chong Wang et al.
Journal of cellular and molecular medicine (2020-12-09)
In this study, we investigated the ability of the Polysaccharide from the Eggs of Strongylocentrotus nudus (SEP) to regulate cellular autophagy and apoptosis in leukaemia cells. Human acute myeloid leukaemia (AML) cells (HL60) and murine AML cells (L1210) treated with
Alexander Lucius et al.
Cellular signalling, 28(3), 136-147 (2015-12-23)
3-Iodothyronamine (3T1AM) is an endogenous thyroid hormone metabolite that interacts with the human trace amine-associated receptor 1 (hTAAR1), a G-protein-coupled receptor, to induce numerous physiological responses including dose-dependent body temperature lowering in rodents. 3T1AM also directly activates cold-sensitive transient receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service