I3282
Monoclonal Anti-Interleukin-10 Receptor α antibody produced in mouse
clone 37607, purified immunoglobulin, lyophilized powder
Synonym(s):
Anti-IL-10 Receptor α
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
37607, monoclonal
form
lyophilized powder
species reactivity
human
technique(s)
neutralization: suitable
isotype
IgG1
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... IL10RA(3587)
General description
Interleukin-10 (IL-10) is a pleotropic cytokine with anti-inflammatory and immunosuppressive functions. It is the key cytokine that regulates the host inflammatory responses to bacterial, fungal and viral pathogens. IL-10 is secreted by a variety of cells including T cells, B cells, dendritic cells, eosinophils, natural killer cells and mast cells. The effects of IL-10 are mediated by binding to the IL-10R, consisting of α and β subunits. The signalling through IL-10R activates Jak1, Tyk2 protein kinases and JAK/STAT3 pathways. IL-10R is involved in suppression of innate immunity to kill pathogens and inhibits the release of reactive oxygen and nitrogen species from activated macrophages. Regulation of IL-10R mediated signalling is important in infections, inflammatory responses in autoimmune encephalomyelitis, endotoxic shock and chronic enterocolitis
Monoclonal Anti-Interleukin-10 Receptor α (IL-10 Rα) recognizes human IL-10 Rα. The antibody does not cross-react with recombinant mouse IL-10 Ra.
Monoclonal Anti-Interleukin-10 Receptor α (IL-10 Rα) recognizes human IL-10 Rα. The antibody does not cross-react with recombinant mouse IL-10 Ra.
Specificity
The antibody has the ability to block cell surface human IL-10 Rα mediated bioactivity induced by recombinant human IL-10. The antibody does not cross-react with mouse IL-10 Rα.
Immunogen
purified recombinant human IL-10 soluble receptor α expressed in insect Sf 21 cells.
Application
Anti-interleukin-10 receptor α antibody is suitable for neutralization reactions (ND50 is 0.03-0.1 μg/ml).
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yan Zhang et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 30(3), 113-122 (2010-03-02)
Apoptosis and the rapid clearance of apoptotic cells (ACs) by professional or nonprofessional phagocytes are normal and coordinated processes that ensure controlled cell growth and stress response with nonpathological outcomes. Uptake of ACs by phagocytes is thought to suppress autoimmune
Christian M Hedrich et al.
Immunologic research, 47(1-3), 185-206 (2010-01-21)
IL-10 plays an essential part in controlling inflammation and instructing adaptive immune responses. Consequently, dysregulation of IL-10 is linked with susceptibility to numerous infectious and autoimmune diseases in mouse models and in humans. It has become increasingly clear that appropriate
Joshua C Cyktor et al.
Infection and immunity, 79(8), 2964-2973 (2011-05-18)
The generation of an effective immune response against an infection while also limiting tissue damage requires a delicate balance between pro- and anti-inflammatory responses. Interleukin-10 (IL-10) has potent immunosuppressive effects and is essential for regulation of immune responses. However, the
K W Moore et al.
Annual review of immunology, 19, 683-765 (2001-03-13)
Interleukin-10 (IL-10), first recognized for its ability to inhibit activation and effector function of T cells, monocytes, and macrophages, is a multifunctional cytokine with diverse effects on most hemopoietic cell types. The principal routine function of IL-10 appears to be
Ivan J Fuss et al.
Journal of immunology (Baltimore, Md. : 1950), 168(2), 900-908 (2002-01-05)
In the present study, we define the relation between TGF-beta and IL-10 in the regulation of the Th1-mediated inflammation occurring in trinitrobenzene sulfonic acid (TNBS)-colitis. In initial studies, we showed that the feeding of trinitrophenol-haptenated colonic protein to SJL/J mice
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service