Skip to Content
Merck
All Photos(4)

Key Documents

I2255

Sigma-Aldrich

Inulin from chicory

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (chicory)

form

powder

impurities

≤0.05% free glucose (enzymatic assay)
<15% water (Karl Fischer)

color

white

mp

158 - 165 °C ((316 - 329 °F ))

solubility

water: 100 mg/mL, clear to hazy, colorless

storage temp.

room temp

SMILES string

OC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O.OC[C@H]2OC(O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O

InChI

1S/C12H22O11.C6H12O6/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12;7-1-3-4(9)5(10)6(11,2-8)12-3/h4-11,13-20H,1-3H2;3-5,7-11H,1-2H2/t4-,5-,6-,7-,8+,9-,10+,11?,12+;3-,4-,5+,6-/m11/s1

InChI key

TUULCWHRUVPVEB-UGWHTDGHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potential replacement for high fructose corn syrup due to its prebiotic function and similar structural characteristics to corn syrup.

Biochem/physiol Actions

A heterogeneous blend of fructose polymers that function as plant storage carbohydrates.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marcel B Roberfroid
The Journal of nutrition, 137(11 Suppl), 2493S-2502S (2007-10-24)
A food (ingredient) is regarded as functional if it is satisfactorily demonstrated to affect beneficially 1 or more target functions in the body beyond adequate nutritional effects. The term inulin-type fructans covers all beta(2<--1) linear fructans including native inulin (DP
Pamela I Aldrete-Herrera et al.
Foods (Basel, Switzerland), 8(9) (2019-09-25)
In this study, we characterize fructan extracts from five wild agave varieties at three ages to identify their potential use in the food industry. Physicochemical parameters (solids soluble total and pH), sugar content and fructan distribution profiles by high-performance anion-exchange
Chien-Ning Hsu et al.
Nutrients, 11(12) (2019-12-08)
Hypertension can originate in early life caused by perinatal high-fat (HF) consumption. Gut microbiota and their metabolites short chain fatty acids (SCFAs), trimethylamine (TMA), and trimethylamine N-oxide (TMAO) are involved in the development of hypertension. Despite the beneficial effects of
The Glycolytic Versatility of
Alfonso Benítez-Páez et al.
Frontiers in cellular and infection microbiology, 7, 383-383 (2017-10-04)
Rebeca Cerezuela et al.
Fish & shellfish immunology, 34(3), 843-848 (2013-01-16)
The present work describes effects of dietary inulin, two microalgae (Tetraselmis chuii and Phaeodactylum tricornutum) and Bacillus subtilis (solely or combined with inulin or microalgae) on the expression of different genes in the intestine of the gilthead seabream (Sparus aurata L.)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service