Skip to Content
Merck
All Photos(2)

Key Documents

I0523

Sigma-Aldrich

IL-2 mouse

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Interleukin-2, T-cell growth factor, mIL-2, IL-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

mouse

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

≤0.2 ng/mL ED50

quality

endotoxin tested

mol wt

17.2 kDa

packaging

pkg of 5X20 μg
pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1.000 EU/μg

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il2(16183)

Looking for similar products? Visit Product Comparison Guide

General description

IL-2 (interleukin-2) codes for a pro-inflammatory cytokine. IL-2 is mostly expressed in T helper cells.

Application

Interleukin-2 from mouse has been used to induce sepsis in rat model. It has also been used in dendritic epidermal T cell culture.

Biochem/physiol Actions

Interleukin-2 (IL-2) is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lymphokine promotes long-term growth of activated T cells and related cell types. IL-2 plays a role in the activation and proliferation of natural killer cells, induces γ-interferon and B cell growth factor secretion, and modulates the expression of the IL-2 receptor. IL-2 expression is known to reduce with age and results in influenza infection, neoplasms and lethal autoimmunity. IL-2 participates in T cell proliferation and differentiation and controls the homeostasis of T cell. As a part of immune response, IL-2 stimulates the action of T helper cell 1 and 2 along with endothelial white blood cells of skeletal muscles. IL-2 regulation is controlled by c-rel protein. IL-2 is a neurotrophic molecule and has special functions in cortical, hippocampal, septal, cerebellar and retinal neurons.

Physical form

Lyophilized from a 0.2 μm filtered solution in 10 mM sodium citrate, pH 4.0.

Analysis Note

Activity was determined by dose-dependent stimulation of murine CTLL-2 cells. ED50 : ≤ 0.2 ng/ml (corresponds to a specific activity of 5 × 106 units/mg)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein kinases JAK and ERK mediate protective effect of interleukin-2 upon ganglion cells of the developing rat retina
Marra C, et al.
Journal of Neuroimmunology, 233(1-2), 120-126 (2011)
Decreased c-rel activation contributes to aberrant interleukin-2 expression in CD4+ T cells of aged rats
Gong Z, et al.
Molecular Immunology, 61, 1-6 (2014)
Acute exhaustive exercise regulates IL-2, IL-4 and MyoD in skeletal muscle but not adipose tissue in rats
Neto JCR, et al.
Lipids in Health and Disease, 10, 97-97 (2011)
Krithika Badarinath et al.
Methods in molecular biology (Clifton, N.J.), 1879, 285-297 (2018-06-14)
The interplay of immune cells and stem cells in maintaining skin homeostasis and repair is an exciting new frontier in cutaneous biology. With the growing appreciation of the importance of this new crosstalk comes the requirement of methods to interrogate
A rat model of early sepsis: relationships between gentamicin pharmacokinetics and systemic and renal effects of bacterial lipopolysaccharide combined with interleukin-2
Studena S, et al.
Biological & Pharmaceutical Bulletin, 35(10), 1703-1710 (2012)

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service