Skip to Content
Merck
All Photos(1)

Key Documents

I0138

Sigma-Aldrich

Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM) antibody produced in mouse

clone RG-16, ascites fluid

Synonym(s):

Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

RG-16, monoclonal

contains

15 mM sodium azide

should not react with

pig, chicken, bovine, horse, human, guinea pig, turkey, rat, canine, goat, feline, sheep

technique(s)

indirect ELISA: 1:10,000

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Monoclonal Anti-Rabbit Immunoglobulins (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Immunoglobulins are the members of the eponymous immunoglobulin super-family (IgSF). Immunoglobulins are heterodimeric proteins characterized with two heavy (H) and two light (L) chains. Based on heavy chain C domains immunoglobulins are classified in to five main classes namely IgM, IgG, IgA, IgD, and IgE isotypes.
Reacts with an epitope on the heavy chain of rabbit IgG, IgA and IgM, which is sensitive to reduction. No cross-reactivity with human IgG, IgA and IgM.

Specificity

Binds only rabbit Igs. Does not react with reduced rabbit Igs.

Application

Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM) antibody produced in mouse has been used in enzyme linked immunosorbent assay (ELISA), immunoblots, dot blots and immunohistochemistry.
The mouse monoclonal anti-Rabbit IgG (heavy chain-specific) peroxidase conjugated antibody can be used as a secondary antibody for western blot analysis of protein preps from E. coli cultures at a concentration of 1:10000 diluted in PBSt for 1 hour at room temperature.

Biochem/physiol Actions

Immunoglobulins acts as a cell-surface receptor for antigen that allows cell signaling and cell activation. In addition, it also acts as a soluble effector molecule that can recognize, bind and neutralize antigens. IgG contributes to immune response including neutralization of toxins and viruses. IgM provides first line of defense and is involved in immunoregulation. IgA protects mucous membranes against toxins, virus and bacteria by direct neutralization or by prevention of binding to the mucosal membranes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Torsten Sacher et al.
Cell host & microbe, 3(4), 263-272 (2008-04-15)
The course of systemic viral infections is determined by the virus productivity of infected cell types and the efficiency of virus dissemination throughout the host. Here, we used a cell-type-specific virus labeling system to quantitatively track virus progeny during murine
Li Ji et al.
Experimental and therapeutic medicine, 13(2), 551-559 (2017-03-30)
Nuclear receptor related-1 protein (Nurr1) is a novel orphan member of the nuclear receptor superfamily (the NR4A family) involved in tumorigenesis. The aim of the present study was to investigate the expression and possible function of Nurr1 in pancreatic ductal
Orphan nuclear receptor Nurr1 as a potential novel marker for progression in human pancreatic ductal adenocarcinoma
Ji L, et al.
Experimental and Therapeutic Medicine, 13(2), 551-559 (2017)
Cayla A Wood et al.
Nature communications, 12(1), 5410-5410 (2021-09-15)
Photoacoustic (PA) imaging is a functional and molecular imaging technique capable of high sensitivity and spatiotemporal resolution at depth. Widespread use of PA imaging, however, is limited by currently available contrast agents, which either lack PA-signal-generation ability for deep imaging
Sangheon Han et al.
Biomedical optics express, 10(7), 3472-3483 (2019-07-31)
Gold nanoparticles (AuNPs) below 10 nm in size can undergo renal clearance, which could facilitate their clinical translation. However, due to non-linear, direct relationship between their absorption and size, use of such "ultra-small" AuNPs as contrast agents for photoacoustic imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service