Skip to Content
Merck
All Photos(1)

Documents

HT20

Sigma-Aldrich

Iron Stain

Synonym(s):

Prussian Blue Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Principle

Deposits of loosely bound iron in erythrocytes, bone marrow, or tissue stain an intense blue when treated with acid ferrocyanide according to the well-known Prussian blue reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Venugopal et al.
Scientific reports, 10(1), 16514-16514 (2020-10-07)
Proprotein convertase subtilisin/kexin type 9 (PCSK9) deficiency leads to lower cholesterol and is associated with reduced vascular complications in the general population. Cholesterol lowering may also have beneficial effects in sickle cell disease (SCD). The objective of this study was
Jessica Cooperrider et al.
Experimental neurology, 277, 27-34 (2015-12-25)
The olivo-cerebellar system has an essential role in the detection and adaptive correction of movement errors. While there is evidence of an error signal in the cerebellar cortex and inferior olivary nucleus, the deep cerebellar nuclei have been less thoroughly
Carl Maximilian Thielmann et al.
Scientific reports, 9(1), 11326-11326 (2019-08-07)
Iron-loaded tumor-associated macrophages (iTAMs) show a pro-inflammatory phenotype, hallmarked by anti-tumorigenic activity and an ability to attenuate tumor growth. Here we explored the relevance of these findings in lung cancer patients by investigating the impact of the iTAM content in
Di Ding et al.
Metabolic brain disease, 35(2), 275-282 (2019-12-12)
Dysregulation of iron metabolism and aberrant iron deposition has been associated with multiple sclerosis. However, the factors that contribute to this pathological state remain to be understood. In this study, human multiple sclerosis and mice brain samples were analyzed through
Renu John et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(18), 8085-8090 (2010-04-21)
Dynamic magnetomotion of magnetic nanoparticles (MNPs) detected with magnetomotive optical coherence tomography (MM-OCT) represents a new methodology for contrast enhancement and therapeutic interventions in molecular imaging. In this study, we demonstrate in vivo imaging of dynamic functionalized iron oxide MNPs

Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service