Skip to Content
Merck
All Photos(2)

Key Documents

HS24319

Pop-Up tube rack

18 x 50 mL, green polypropylene rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.17
Pricing and availability is not currently available.

material

green polypropylene rack

feature

18 x 50 mL

packaging

pack of 2 ea

manufacturer/tradename

Heathrow Scientific HS24319

L × W × H

255 mm × 137 mm × 72 mm

Looking for similar products? Visit Product Comparison Guide

General description

Simple, easy action to open and close - expand and collapse when racks are needed. Designed to help organize 15 and 50 mL tubes. Imprinted grid aides quick retrieval. Stack racks in open or closed position. Saves space when racks are not in use! Robust polypropylene construction. Not recommended for cleaning in autoclave.

Measures 255 x 137 x 72 mm when expanded; 21 mm H when collapsed.
The Pop-Up Rack collapses for compact storage and “pops-up” for quick use, easy open and close.

Features and Benefits

  • Constructed of polypropylene
  • Easy to open and close
  • Alphanumeric Identification for quick location and retrieval of tubes
  • Stack racks in open or closed position
  • Saves space when racks are not in use
  • Not recommended for autoclaving

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service