Key Documents
HS24311A
Peg Tube Rack
to hold, 50 tube(s) 14-17 mm tubes, white
About This Item
UNSPSC Code:
41122800
NACRES:
NB.17
material
fiberglass
polypropylene (reinforced)
white
feature
to hold 50 tube(s) 14-17 mm tubes
packaging
pack of 2 ea
manufacturer/tradename
Heathrow Scientific HS24311A
L × W × H
188 mm × 104 mm × 69 mm
Related Categories
General description
The Peg Tube Rack is strong, tough, and versatile for bench or tube storage. It is constructed of fiberglass-reinforced polypropylene, which is ideal for bench work or tube storage.
Features and Benefits
  • Constructed of fiberglass-reinforced polypropylene
  • Spacious peg design allows easy viewing of tube contents
  • Use as a drying rack for glass plates or upside-down tubes
  • Pegs remain straight even after repeated autoclaving
  • Embossed alphanumeric identification for quick location and retrieval of tubes
  • Unique tear drop well design allows for better retention of tubes
  • Autoclavable
Choose from one of the most recent versions:
Certificates of Analysis (COA)