Skip to Content
Merck
All Photos(7)

Key Documents

HPA054116

Sigma-Aldrich

Anti-TLE3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ESG, Anti-ESG3, Anti-GRG3, Anti-HsT18976, Anti-KIAA1547

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GSQAHLTVKDEKNHHELDHRERESSANNSVSPSESLRASEKHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLE3(7090)

General description

Transducin-like enhancer of split 3 (TLE3) belongs to the groucho family and comprises Q and WD40 domains. The TLE3 gene is mapped to human chromosome locus 15q23. It is expressed in adipocytes and shows higher levels in white adipose tissue (WAT) than the brown adipose tissue (BAT). It is also expressed in the prostate. TLE3 exists in isoforms due to alternative splice events.

Immunogen

transducin-like enhancer of split 3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Transducin-like enhancer of split 3 (TLE3) acts as a modulator of adipogenesis. It is a transcriptional integrator for the nuclear receptor, peroxisome proliferator-activated receptor γ (PPARγ). TLE3 also interacts with Notch and controls the differentiation of epithelial cells. It serves as a transcriptional repressor for adenomatous polyposis coli (APC) and β-catenin in the wingless (Wnt) pathway by its interaction with the chromatin complex. TLE3 mediates inhibition of mitochondrial gene expression and early B-cell factor (EBF2) in beige adipocytes and regulates thermogenic gene expression. It may serve as a therapeutic target for obesity and diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85538

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Goli Samimi et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 21(2), 273-279 (2011-12-24)
We have previously shown that transducin-like enhancer of split 3 (TLE3) is associated with outcome specifically in patients with taxane-treated breast cancer and not in patients treated with anthracycline-based regimens without a taxane. The purpose of this study was to
Helder I Nakaya et al.
Biochemical and biophysical research communications, 364(4), 918-923 (2008-02-15)
The TLE genes constitute a family of important transcriptional co-repressors involved in many cellular processes. We found evidence of alternatively spliced mRNAs for human TLE1-4 containing premature stop codons, thus encoding putative shortened proteins. Microarray experiments and Real-time RT-PCR assays
Valéria Tavares et al.
Data in brief, 30, 105399-105399 (2020-04-08)
Venous thromboembolism (VTE) is a common cardiovascular disease, for which several single nucleotide polymorphisms (SNPs) underlying susceptibility were identified. Apart from candidate gene approach, genome-wide association studies (GWAS) have contributed to the identification of novel VTE-associated SNPs, including some with
Claudio J Villanueva et al.
Cell metabolism, 13(4), 413-427 (2011-04-05)
PPARγ and Wnt signaling are central positive and negative regulators of adipogenesis, respectively. Here we identify the groucho family member TLE3 as a transcriptional integrator of the PPARγ and Wnt pathways. TLE3 is a direct target of PPARγ that participates
Stephanie Pearson et al.
Genes & development, 33(13-14), 747-762 (2019-05-28)
Prolonged cold exposure stimulates the recruitment of beige adipocytes within white adipose tissue. Beige adipocytes depend on mitochondrial oxidative phosphorylation to drive thermogenesis. The transcriptional mechanisms that promote remodeling in adipose tissue during the cold are not well understood. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service