Skip to Content
Merck
All Photos(4)

Key Documents

HPA049838

Sigma-Aldrich

Anti-PTRF antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAVIN1, Anti-cavin-1, Anti-polymerase I and transcript release factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000
western blot: 0.04-0.4 μg/mL

immunogen sequence

EPSSAGAQAAEEPSGAGSEELIKSDQVNGVLVLSLLDKIIGAVDQIQLTQAQLEERQAEMEGAVQSIQGELSKLGK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTRF(284119)

General description

PTRF (polymerase I and transcript release factor) is a cytoplasmic protein, that is also known as Cav-p60 or Cavin. It is located on chromosome 17q21.2.

Immunogen

polymerase I and transcript release factor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PTRF (polymerase I and transcript release factor) is an essential factor for the formation of caveola. It modulates the structure and function of caveola. Mutations in PTRF cause muscular dystrophy with generalized lipodystrophy. It actively participates in lipid metabolism and gene expression modulated by insulin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83755

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Yan Lim et al.
Cellular microbiology, 19(12) (2017-08-05)
Caveolae are composed of 2 major proteins, caveolin 1 (CAV1) and cavin 1 or polymerase transcript release factor I (CAVIN1). Here, we demonstrate that CAV1 levels modulate invasion of Group A Streptococcus (GAS) into nonphagocytic mammalian cells. GAS showed enhanced
Identification of a novel nonsense mutation and a missense substitution in the AGPAT2 gene causing congenital generalized lipodystrophy type 1.
Haghighi A, et al.
European Journal of Medical Genetics, 55(11), 620-624 (2012)
PTRF-Cavin, a conserved cytoplasmic protein required for caveola formation and function.
Hill MM, et al.
Cell, 132(1), 113-124 (2008)
Human PTRF mutations cause secondary deficiency of caveolins resulting in muscular dystrophy with generalized lipodystrophy.
Hayashi Y K, et al.
The Journal of Clinical Investigation, 119(9), 2623-2633 (2009)
Jin-Yih Low et al.
Molecular cancer research : MCR, 18(9), 1414-1426 (2020-06-05)
Lipid uptake occurs through caveolae, plasma membrane invaginations formed by caveolins (CAV) and caveolae-associated protein 1 (CAVIN1). Genetic alterations of CAV1N1 and CAV1 modify lipid metabolism and underpin lipodystrophy syndromes. Lipids contribute to tumorigenesis by providing fuel to cancer metabolism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service