Skip to Content
Merck
All Photos(8)

Key Documents

HPA036576

Sigma-Aldrich

Anti-TPPP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TPPP/p25, Anti-TPPP1, Anti-p25, Anti-p25alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

AKKRFKDKSSEEAVREVHRLIEGKAPIISGVTKAISSPTVSRLTDTTKFTGSHK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPPP(11076)

General description

Tubulin polymerization promoting protein (TPPP), also known as p25, is expressed in myelinating oligodendrocytes. TPPP is mapped to human chromosome 5p15.33. It has a p25 domain and is a disordered protein.

Immunogen

tubulin polymerization promoting protein

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tubulin polymerization promoting protein (TPPP) regulates oligodendrocyte differentiation. It interacts with tubulinand regulates microtubule bundle formation. Phosphorylation of TPPP at threonine or serine residue by mitogen-activated protein kinase 2 or by LIM kinase (LIMK) impactstubulin polymerization functionality. High expression of TPPP favors acetylation of microtubules. Deletion in the gene locus of TPPP may be implicated in the pathogenesis of cerebellar hypoplasia and facial dysmorphism in fetus. Mutations in the TPPP gene contributes to tumor progression in small cell lung cancer. TPPP localization with α-synuclein may help in the understanding neurodegeneration in Parkinson′s and Alzheimer′s diseases. TPPP promotes abnormal microtubule assemblies in alpha-synucleinopathies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80111

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TPPP/p25 promotes tubulin acetylation by inhibiting histone deacetylase 6
Tokesi N, et al.
The Journal of Biological Chemistry, 285(20), 17896-17906 (2010)
Jacques Gilloteaux et al.
Ultrastructural pathology, 45(2), 128-157 (2021-06-23)
A murine osmotic demyelination syndrome (ODS) model of the central nervous system included the relay thalamic ventral posterolateral (VPL) and ventral posteromedial (VPM) nuclei. Morphologic comparisons between treatments have revealed oligodendrocyte changes and, already 12 hours following the osmolality restoration, some
The phosphorylation of p25/TPPP by LIM kinase 1 inhibits its ability to assemble microtubules
Acevedo K, et al.
Experimental Cell Research, 313(20), 4091-4106 (2007)
P25alpha tubulin polymerization promoting protein expression by myelinating oligodendrocytes of the developing rat brain
Skjoerringe T, et al.
Journal of Neurochemistry, 99(1), 333-342 (2006)
TPPP/p25: from unfolded protein to misfolding disease: prediction and experiments
Orosz F, et al.
Biology of the Cell, 96(9), 701-711 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service