Skip to Content
Merck
All Photos(6)

Key Documents

HPA036087

Sigma-Aldrich

Anti-SLC35A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-UGALT, Anti-UGAT, Anti-UGT, Anti-UGT1, Anti-UGT2, Anti-UGTL, Anti-solute carrier family 35 (UDP-galactose transporter), member A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RGAAKAIASASASASGPCVHQQPPGQPPPPQLSSHRGDLITEPFLPKLLTKVKGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC35A2(7355)

General description

Solute carrier family 35 member A2 (SLC35A2) gene is mapped to human chromosome Xp11.23. This gene codes for Uridine diphosphate (UDP)-galactose transporter (UGT). UGT belongs to the family of nucleotide sugar transporters (NSTs). UGT1 and UGT2 are the two alternative splicing variants of UGT.

Immunogen

solute carrier family 35 (UDP-galactose transporter), member A2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SLC35A2 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Uridine diphosphate (UDP)-galactose transporter (UGT) facilitates the transport of UDP-galactose to the Golgi apparatus. Short N-terminal region of UGT is responsible for galactosylation of N-glycans. Mutation in the gene is associated with the development of congenital disorders of glycosylation (CDG) and early-onset epileptic encephalopathy and developmental delay.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78271

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genome-wide CRISPR screen reconciles the role of N-linked glycosylation in galectin-3 transport to the cell surface
Stewart SE, et al.
Journal of Cell Science, 130(19), 3234-3247 (2017)
Short N-terminal region of UDP-galactose transporter (SLC35A2) is crucial for galactosylation of N-glycans
Sosicka P, et al.
Biochemical and Biophysical Research Communications, 454(4), 486-492 (2014)
A case of early onset epileptic encephalopathy with de novo mutation in SLC35A2: clinical features and treatment for epilepsy
Kimizu T, et al.
Brain & Development, 39(3), 256-260 (2017)
SLC35A2-related congenital disorder of glycosylation: Defining the phenotype
Yates TM, et al.
European Journal of Paediatric Neurology, 22(6), 1095-1102 (2018)
Chien-Liang Liu et al.
In vivo (Athens, Greece), 37(1), 262-269 (2023-01-03)
A recent study suggested that solute carrier family 35 member A2 (SLC35A2) is related to poor prognosis in patients with breast cancer. SLC35A2 transports uridine diphosphate-galactose from the cytosol to the lumen of the endoplasmic reticulum and Golgi. Immunohistochemical expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service