Skip to Content
Merck
All Photos(3)

Key Documents

HPA036070

Sigma-Aldrich

Anti-PTK6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRK, Anti-PTK6 protein tyrosine kinase 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PYPGMSNHEAFLRVDAGYRMPCPLECPPSVHKLMLTCWCRDPEQRPCFKALRERLSSFTSYENPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK6(5753)

General description

PTK6 (protein tyrosine kinase 6) belongs to intracellular tyrosine kinases family. It is normally expressed in epithelial cells of skin, gastrointestinal tract and prostate. It is also called as breast tumor kinase (brk), for its critical role in progression of breast cancer. The PTK6 gene is located on the human chromosome 20q13.33. PTK6 protein is expressed in two isoforms.

Immunogen

PTK6 protein tyrosine kinase 6 recombinant protein epitope signature tag (PrEST)

Application

Anti-PTK6 rabbit polyclonal antibody has been used in microarray analysis of PTK6 in breast tumor samples. It may also be used to detect protein tyrosine kinase 6 using western blot analysis in esophageal squamous cell carcinoma.

Biochem/physiol Actions

Protein tyrosine kinase 6 regulates epithelial cell gene expression by modulating RNA binding proteins. It favours epidermal growth factor receptor signalling cascade. Mutations in kinase domain leads to activity suppression. PTK6 is effectively inhibited by tumor suppressor protein phosphatase and tensin homolog (PTEN) in human prostate cancer tissues and cell lines. PTK6 is highly expressed and up-regulated in a multitude of human carcinomas including hepatocellular carcinoma, cervical squamous carcinoma, prostate cancer and colon cancer. PTK6 mediates proteasome-mediated degradation of tumor suppressor protein by phosphorylation. With the advent of its association in human cancer, PTK6 is considered as a potential drug target for cancer chemotherapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78020

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTK6 promotes hepatocellular carcinoma cell proliferation and invasion
Chen X, et al.
American Journal of Translational Research, 8(10), 4354-4354 (2016)
Protein tyrosine kinase 6 negatively regulates growth and promotes enterocyte differentiation in the small intestine
Haegebarth A, et al.
Molecular and Cellular Biology, 26(13), 4949-4957 (2006)
PTK6 inhibition suppresses metastases of triple-negative breast cancer via SNAIL-dependent E-cadherin regulation
Ito K, et al.
Cancer Research, 76(15), 4406-4417 (2016)
The nuclear tyrosine kinase BRK/Sik phosphorylates and inhibits the RNA-binding activities of the Sam68-like mammalian proteins SLM-1 and SLM-2
Haegebarth A, et al.
The Journal of Biological Chemistry, 279(52), 54398-54404 (2004)
BRK/Sik expression in the gastrointestinal tract and in colon tumors
Llor X, et al.
Clinical Cancer Research, 5(7), 1767-1777 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service